Ca/CaMKs
Free calcium is a major second messenger in all cell types. One mechanism by which calcium ions exert their effects is by binding to a 17 kDa protein, calmodulin (CaM). The binding of four calcium ions to calmodulin changes its conformation and promotes its interaction with a number of other proteins, including several classes of protein kinases that are activated by the calcium/CaM complex. A practical way of classifying the calcium/CaM-dependent protein kinases is based on their substrate specificity: some of these enzymes have only one substrate, and are designated as 'dedicated' calcium/CaM-dependent protein kinases, while others have broad substrate specificity and are termed 'multifunctional' kinases.
The dedicated calcium/CaM-dependent protein kinases comprise three enzymes: phosphorylase kinase, myosin light chain kinase and eEF2-kinase. Phosphorylase kinase, the first protein kinase to be identified, phosphorylates and activates glycogen phosphorylase, the enzyme that degrades glycogen. Phosphorylase kinase is activated either by phosphorylation by cAMP-dependent protein kinase or by binding of calcium/CaM. This mechanism of regulation is especially important in muscle where glycogen breakdown and muscle contraction are coordinated by the transient increase in cytosolic calcium levels. Myosin light chain kinases (MLCK) are a group of enzymes that phosphorylate the regulatory light chain of myosin. MLCK induces smooth muscle contraction by increasing actin-activated myosin ATPase activity. In contrast, striated muscle MLCK plays only a modulatory role in contraction by potentiating the effects of troponin-bound calcium on actin/myosin. In non-muscle cells, MLCKs are key factors in the numerous processes which involve actin/myosin-based organelle movement or cell motility. eEF2-kinase (previously known as CaM-kinase III) phosphorylates eukaryotic elongation factor 2 (eEF2), a GTPase necessary for the elongation step in protein translation. eEF2-kinase belongs to a separate class of protein kinases, the α kinases, with no sequence similarity with the main family of protein kinases. Phosphorylation of eEF2 by eEF2-kinase accounts for a calcium-dependent interruption of protein synthesis that may be responsible for a rapid change in the nature of the mRNA being translated.
Multifunctional calcium/CaM-dependent protein kinases comprise three enzymes referred to as CaM-kinases I, II and IV. CaM-kinase II (CaMKII) is an oligomer of 12 subunits, which has unique properties and is also the most extensively studied. It is a ubiquitously distributed enzyme highly enriched in neurons, especially in post-synaptic densities. As is the case of other CaM-kinases, the activity of CaMKII is inhibited by an autoinhibitory domain. This inhibition is alleviated by binding of calcium/CaM which allows autophosphorylation of the autoinhibitory domain. Once autophosphorylation has occurred, the presence of calcium/CaM is no longer necessary and the enzyme becomes calcium/CaM-independent. Interestingly, the oligomeric structure of CaMKII and the fact that autophosphorylation is a 'trans' reaction between different subunits of the oligomer has important consequences. Autophosphorylation promotes calcium/CaM trapping and occurs only when two adjacent subunits are bound to calcium/CaM. Thus, CaMKII is sensitive to the duration and frequency of calcium transients, and therefore is capable of decoding the frequency of calcium spikes. CaMKII may also remain active for some time while calcium levels return to normal, thereby maintaining a transient 'memory' of neuronal activation. CaMKII is a very important contributor to the processes of synaptic plasticity and LTP induction.
CaMKI and CaMKIV are monomeric enzymes that share the common property of being activated by calcium/CaM binding and by phosphorylation by a CaM-kinase-kinase (CaMKK). Thus, together these kinases are organized as a calcium/CaM-dependent protein kinase cascade. CaMKI is a ubiquitously expressed largely cytosolic enzyme which phosphorylates many substrates, including synapsin I. In contrast, CaMKIV (also known as CaMK-Gr because of its abundance in cerebellar granule cells) is located in the nucleus. CaMKIV phosphorylates transcription factors, including cAMP responsive element binding protein (CREB) and the associated CREB-binding protein (CBP), and thus plays a major role in calcium-regulated gene transcription. CaMKK controls the activity of both CaMKI and CaMKIV. There are two isoforms of CaMKK, α and β, enriched in the cytoplasm and the nucleus, respectively. CaMKK is also able to phosphorylate and activate PKB, and thus exert anti-apoptotic effects. A family of pro-apoptotic serine/threonine protein kinases has been identified and termed Death Associated Protein Kinases (DAP-kinases). Two of these kinases possess a CaM-binding domain and are activated by calcium-CaM.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Family Members CaMKK DAP-K Phosphorylase Kinase (P2014) MLCK
Other Names Calcium/calmodulin-dependent
protein kinase kinase
Death-associated protein kinase PHK
Glycogen phosphorylase kinase, GPK (P6635)
Myosin light chain kinase
Molecular Weight
(kDa)
60-70 kDa 165 kDa γ : 45 kDa 70-150 kDa
Structural Data Monomer Monomer 16 subunits (α4, β4, γ4, δ4) Monomer
Isoforms α, β One
Several shorter related kinases
Two catalytic (M muscle, L/T, liver/testis) Several
Species Mammals
Birds
Vertebrates Vertebrates
Drosophila
Vertebrates
Drosophila
Domain
Organization
Catalytic
CaM-R
Catalytic
CaM-R
ankyrin repaeats
cytoskeleton binding
death domain
γ
Catalytic
CaM-R
Actin binding
PEVK repeats
Ig
FN
Catalytic
CaM-R
Phosphorylation
Sites
Not characterized Ser308
(autophos., inhibition)
Ser27 in β subunit
(PKA, activation)
Thr803
(autophos., Ca2+/CaM-independence)
Ser815 and Ser823
(autophos., inhibition)
Tissue
Distribution
Ubiquitous
Brain
Testis
Spleen
Ubiquitous Ubiquitous
Liver
Muscle
Ubiquitous
Subcellular
Localization
Cytoplasm, nucleus Cytoplasm Cytoplasm Cytoplasm
Binding Partners/
Associated Proteins
Not Known
Actin filaments Not Known
Actin (A3653, A2522)
Upstream
Activators
Ca2+/calmodulin Ca2+/calmodulin
Dephosphorylation
Ca2+/calmodulin
PKA (P2645)
ADP (A2754)
Ca2+/calmodulin
ERK (E9402)
Downstream
Activation
CaMKI
CaMKIV
PKB (A8729)
AMP-kinase (P6998)
Myosin light chain Glycogen phosphorylase Myosin light chain
Activators Not Known
Not Known
Not Known
Not Known
Inhibitors STO-609 (S1318) K252A (K1639)
MW01-026Z
3-amino-6-phenyl- pyridazine
Not Known
ML-7 (I2764)
ML-9 (C1172)
Selective
Activators
Not Known
Not Known
Not Known
Not Known
Physiological
Function
Activates CaMKI, IV, cell survival Membrane blebbing Promotes glycogen degradation Smooth muscle contraction
Disease
Relevance
Not Known
Apoptosis
Neuronal death
Mutations in glycogenosis Vasospasm
Asthma
Family Members eEF2-Kinase CaMKI CaMKII
CaMKIV
Other Names Eukaryotic elongation factor-2 kinase
Cam kinase III
Calcium/calmodulin-dependent
protein kinase 1
Calcium/calmodulin-dependent
protein kinase 2
Calcium/calmodulin-dependent
protein kinase 4
Molecular Weight
(kDa)
~100 kDa ~42 kDa 50-60 kDa 65-67 kDa
Structural Data Monomer
Unrelated to classical kinases
Monomer Oligomer (12) Monomer
Isoforms One α
β
γ
α
β
γ
δ
Not Known
Species Vertebrates Vertebrates
Drosophila
Vertebrates
Drosophila
Mammals
Domain
Organization
N-terminal α-kinase d Catalytic
CaM-R
Oligomerization domain
catalytic
CaM-R
Catalytic
CaM-R
Phosphorylation
Sites
Ser359  (p38-MAPK)
Ser366 (p90RSK)
Ser377 (MAPKAP-K2)
Ser500 (PKA)
Thr177
(CaMKK, activation)
Autophosphorylation (CaM-R)
others
CaMKK
(Thr196, activation loop)
Tissue
Distribution
Ubiquitous Ubiquitous Ubiquitous Neurons
Testis
T cells
Subcellular
Localization
Cytoplasm Cytoplasm Cytoplasm
Nucleus
Cytoplasm
Nucleus
Binding Partners/
Associated Proteins
Not Known
Not Known
Not Known
Not Known
Upstream
Activators
Ca2+/calmodulin Ca2+/calmodulin
CaMKK
Ca2+/calmodulin
autophosphorylation
Ca2+/calmodulin
CaMKK
Downstream
Activation
eEF-2 Multiple
(e.g. synapsin I, CREB)
Multiple
(e.g. NMDA and AMPA receptors)
Multiple
(e.g. MEF2)
Activators Not Known
Not Known
Not Known
Not Known
Inhibitors TS-2
TS-4
KN-62 (I2142)
KN-93 (K1385)
KN-62 (I2142)
KN-93 (K1385)
KN-62 (I2142)
KN-93 (K1385)
Selective
Activators
Not Known
Not Known
Not Known
Not Known
Physiological
Function
Protein synthesis interruption Many
(e.g. transcription regulation)
Many
(e.g. synaptic plasticity)
Many
(e.g. transcription regulation)
Disease
Relevance
Not Known
Not Known
Not Known
Not Known
Abbreviations
CaMK: Calcium/Calmodulin-Dependent Protein Kinase
CaMKK: Calcium/Calmodulin-Dependent Protein Kinase Kinase
CaM-R: Calmodulin regulatory domain: binds calmodulin and regulates kinase activity
CREB: cAMP-response element-binding protein
DAP-kinase: Death-Associated Protein kinase
eEF2: eukaryotic Elongation Factor 2
FN: fibronectin domain
Ig: immunoglobulin domain
KN-62: 2-[N-(4’-Methoxybenzenesulfonyl)]amino-N-(4’-chlorophenyl)-2-propenyl-N-methylbenzylamine phosphate
KN-93: N-(2-[N-[4-Chlorocinnamyl]-N-methylaminomethyl]phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide
ML-7: 1-(5-Iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine
ML-9: 1-(5-Chloronaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine
MAPK: mitogen-activated protein kinase
MAPKAP-K: MAPK-activated protein kinase
MEF2: Myocyte Enhancer Factor-2
MLCK: Myosin Light Chain Kinase
PKA: cAMP-dependent protein kinase
PKB: Protein Kinase B (also known as Akt)
RSK: ribosome subunit S6 kinase
TS-2: 4-Ethyl-4-hydroxy-2-p-tolyl-5,6-dihydro-4H-1,3-selenazine
TS-4: 4-Hydroxy-6-isopropyl-4-methyl-2-p-tolyl-5,6-dihydro-4H-1,3-selenazine
Similar Products
References