Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Fak
The focal adhesion kinase (FAK) is a cytoplasmic protein tyrosine kinase that distinctly co-localizes with integrins at sites of attachment to their ligands. In cells in culture, these sites are manifested as regions of close contact with the underlying substrate called focal adhesions. Attachment of integrins to their extracellular matrix ligands is a major regulatory stimulus for FAK, resulting in its tyrosine phosphorylation and enzymatic activation. Other stimuli, e.g. growth factors, neuropeptides, cytokines, mechanical stimuli, can also induce FAK phosphorylation/activation. FAK associates with a large number of enzymes, adaptor and scaffold proteins and serves both enzymatic and scaffolding roles in the transduction of signals.
FAK is organized into 4 domains. At the N-terminus is a FERM domain, which is found in a number of cytoskeletal and signaling proteins and functions to mediate protein-protein interactions. The central region of FAK contains the catalytic domain. The C-terminal domain of FAK contains two distinct regions. The C-terminal 140 amino acids comprise the focal adhesion targeting (FAT) domain, a four α-helix bundle containing binding sites for paxillin, and functions to localize FAK to focal adhesions. Between the catalytic and FAT domains is a region of undefined structure, containing two proline-rich regions that serve as ligands for the SH3 domains of several signaling proteins.
Upon activation, FAK autophosphorylates creating a docking site for the SH2 domains of a number of signaling molecules, including Src family kinases and phosphatidyl-inositol 3’-kinase. Src family kinases then promote phosphorylation of FAK on several other tyrosine residues, resulting in maximal FAK catalytic activity and creation of additional binding sites for other proteins. FAK associates with several other proteins that are tyrosine phosphorylated following integrin-dependent adhesion, p130cas and paxillin, and FAK promotes phosphorylation of these substrates.
FAK is an essential gene in the mouse. FAK has been implicated as a downstream signaling molecule that functions in the control of several integrin regulated biological processes, including cell migration, cell survival and cell proliferation. Recent studies have further defined the role of FAK and these cellular functions in a broader biological context. For example, FAK has been implicated in the control of tubule formation by endothelial cells and angiogenesis under certain circumstances in an animal model. Interesting findings also suggest that FAK may function in the control of neurite outgrowth and netrin induced axonal guidance. Dysregulation of motility, survival and proliferation is a hallmark of a number of human pathological conditions, e.g. cancer. Aberrant FAK signaling results in altered cellular phenotypes, including increased invasion, growth in soft agar, tumorigenicity and metastasis. Further, FAK is overexpressed in a number of human cancers, suggesting that FAK may play a role in the pathology of this disease.
Pyk2 is a FAK-related kinase sharing the same overall domain structure and approximately 45% sequence identity. In contrast to FAK, which is ubiquitously expressed, Pyk2 is more restricted in its expression, predominantly in epithelial cells, hematopoietic cells and neural tissue. Pyk2 is a nonessential gene as knockout mice are viable. A number of common stimuli, including growth factors, cytokines and cell adhesion regulate FAK and Pyk2. In general, FAK is more strongly activated by cell adhesion whereas Pyk2 is more strongly activated by soluble ligands. Notably, ligands that stimulate elevation of cytoplasmic calcium activate Pyk2. There are a number of common binding partners for FAK and Pyk2, e.g. Src family kinases and paxillin, suggesting some common signaling mechanisms. On the other hand, there are FAK specific ligands, e.g. DCC, and Pyk2 specific ligands, e.g. gelsolin, which play roles in distinct functions of the two kinases. Pyk2 may play important roles in macrophage and osteoclast function.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Family Members FAK Pyk2
Other Names FAK56 (D. melanogaster homolog of FAK/Pyk2)
Ptk2 (mouse genomic nomenclature)
p125FAK
CAKβ
CADTK
RAFTK
FAK2
Ptk2b
Molecular Weight 119 kDa (FAKI)
115 kDa
Structural
Information
1052 aa (human, mouse)
1055 aa (rat)
1053 aa (avian)
1009 aa (human, mouse, rat)
Isoforms FAK+ (insert 3 aa after aa 903)
FAK+28, 7 (insert 28 aa after aa 391 and 8 aa after aa 433)
FAK+28, 6, 7 (insert 6 aa after 28 aa insert of FAK+28,7)
FRNK (encodes aa 693-1052)
Pyk2s (aa 739-780 deleted)
PRNK (encodes aa 781-1009)
Species Human
Mouse
Rat
Chicken
Frog
Human
Mouse
Rat
Domain
Organization
Proline-rich region
T-Fak paxillin binding sequence contains talin binding sequence
N-terminal domains bind integrin β subunit
Ferm domains
Phosphorylation
Sites
Tyr397
Tyr407
Tyr576
Tyr577
Tyr861
Tyr925
Ser722
Ser840
Ser843
Ser910
Tyr402
Tyr579
Tyr580
Tyr881
Tissue
Distribution
Ubiquitous
All organs
Lymphoid tissue
Brain
Some fibroblasts
Epithelial cells
Brain
Hematopoietic cells
Subcellular
Localization
Focal adhesions
Cytoplasm
In a few cases in focal adhesions or along stress
fibers, most often diffusely cytosolic
Binding Partners/
Associated Proteins
Src family kinases
PI3K
PLCγ
Grb7
Shc
Grb2
SOCS
p130CAS
HEF1
GRAF
ASAP1
Paxillin and related proteins
Neogenin
DCC
Ezrin
Trio
Growth factor receptors
Etk
EphA1
PIAS1
Integrins
FIP200
Talin
p190RhoGEF
Calpain
Src family kinases
ASAP1
p130CAS
HEF1
PRAP
Pap
Paxillin and related proteins
Gelsolin
Nir family or proteins
FIP200
Upstream
Activators
Integrin-dependent cell adhesion
Growth factors
Neuropeptides
Mechanical stimuli
Growth factors
Cytokines
Ca2+
Nucleotides
Membrane depolarization
Cell adhesion
Downstream
Activation
CAS
Shc
Grb2
PI3K
Nephorocystin
PRAP
Activators Not Known
Not Known
Inhibitors Not Known
Not Known
Selective
Activators
Not Known
Not Known
Physiological
Functions
Cell motility
Cell survival
Cell proliferation
Uptake of pathogenic bacteria
Regulate actin cytoskeleton (focal adhesions)
Cell motility
Uptake of pathogenic bacteria
Bone resorption
Regulate actin cytoskeleton (osteoclast actin ring)
Disease
Relevance
Cancer Not Known
Similar Products
References