Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
InsR
The insulin receptor belongs to a subfamily of receptor tyrosine kinases that also includes the IGF-1 receptor and an orphan receptor called the insulin receptor-related receptor (IRR). It is a tetrameric protein consisting of two α- and two β-subunits encoded by the IR gene. The two subunits derived from a single chain proreceptor undergo post translational processing including cleavage by a furin-like enzyme and glycosylation, forming a single α-β subunit complex. Two of the α-β dimers are then cross linked by disulfide bonds to form the tetramer.
Ligand (insulin or IGF-1) binding to the α-subunit leads to activation of the kinase activity in the β-subunit. Following this initial activation, phosphorylation of one β-subunit by the other (transphosphorylation), leads to a conformational change and a further increase in activity of the kinase domain. Activation of the tyrosine kinase domain leads to autophosphorylation of tyrosine residues in several regions of the intracellular β-subunit, including Tyr960 in the juxtamembrane region, creating an NPXpY recognition motif for the PTB domain of the IRS proteins, Tyr1146, Tyr1150 and Tyr1151 in the regulatory loop and Tyr1316 and Tyr1322 in the C-terminus.
The α-β heterodimers of the individual insulin, IGF-1 and the IRR receptors can form functional hybrids in which ligand binding to one receptor’s binding site leads to activation of the other receptor in the heterodimer by this transphosphorylation process.
Intracellular substrates of the insulin and IGF-1 receptor tyrosine kinases that have been identified include insulin receptor substrate (IRS) proteins 1-4, Gab-1, p62dok, Cbl, APS and the various isoforms of Shc. Following insulin stimulation, the receptor directly phosphorylates most of these substrates on multiple tyrosine residues. These phosphorylated tyrosines occur in specific sequence motifs, which once phosphorylated serve as ‘docking sites’ for intracellular molecules that contain the SH2 (Src-homology) domain, transmitting the insulin signal downstream. A few proteins that bind to phosphotyrosines in the IRS proteins do not contain known SH2 domains; these include the calcium ATPases SERCA 1 and 2, and the SV40 large T antigen.
Negative regulation of insulin receptor signaling has been demonstrated by the tyrosine phosphatase PTP1B, which dephosphorylates the phosphotyrosine residues in the insulin receptor kinase and also through direct association of the novel PIR domain of the Grb14 adaptor protein with the insulin receptor.
Alterations in the function of the insulin receptor, both genetic and acquired, can lead to several different disease states including insulin resistance, diabetes and growth retardation. Insulin resistance at the level of the receptor may be the result of genetic alterations in receptor expression or structure, secondary changes in receptor activity due to serine phosphorylation, or due to down-regulation of receptor concentration. Insulin resistance is also closely linked to other common health problems, including obesity, polycystic ovarian disease, hyperlipidemia, hypertension and atherosclerosis.
The insulin receptor is widely distributed throughout the body, found in tissues classically regarded as both insulin 'responsive', for example muscle, liver and fat, and 'non-responsive', for example brain and the vascular system. It signals through two major signaling pathways, the IRS/PI 3-kinase pathway and the Ras-MAP kinase pathway, controlling processes including glucose transport, uptake and storage, glycogen synthesis, cell growth and differentiation, protein synthesis and gene expression.
Tissue specific knockouts of the insulin receptor have helped to define the role of the receptor in the classical insulin sensitive tissues and identified novel functions in other tissues. The Muscle specific Insulin Receptor Knockout (MIRKO) mouse model exhibits increased insulin stimulated glucose uptake in the fat, suggesting 'cross-talk' between muscle and fat in insulin resistant states. Fat specific knockout (FIRKO) mice have decreased fat mass, are resistant to diet induced obesity and have an extended lifespan, suggesting an interesting role for the insulin receptor in regulating longevity. The neuron specific insulin receptor knockout (NIRKO) has confirmed the importance of the receptor in brain and highlighted a role for it in appetite regulation.
Defining the key steps that lead to specificity in insulin signaling should offer therapeutic approaches for patients suffering from insulin resistant states, including type 2 diabetes.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Family Members Insulin receptor Insulin-like growth factor I receptor IRR
Other Names Insr
IR
IGFR1
JTK13
Somatomedin receptor
INSRR
Insulin receptor-related receptor
IR-related receptor
IRRR
Molecular Weight α-subunit: 135 kDa
β-subunit: 95 kDa
154 kDa 143 kDa
Structural Data α-subunit: 719 aa
β-subunit: 620 aa
1367 aa 1297 aa
Isoforms Not Known IGF1Ra
IGF1Rb
Not Known
Species Mammals
Fish
C. Elegans
Xenopus
Human
Bovine
Mouse
Pig
Rat
Human
Mouse
Rat
Guinea pig
Domain
Organization
Tetramer of 2α and 2β subunits
β-chain contains kinase domains
2 fibronectin type III-like domains
Tetramer of 2 α and 2 β chains
α chain contains ligand-binding domain
β chain contains kinase domain
3 fibronectin type-III domains
Probable tetramer of 2 α and 2 β chains
α chain contains ligand-binding domain
β chain contains kinase domain
4 fibronectin type-III domains
Phosphorylation
Sites
Tyr1146
Tyr150
Tyr1151
Ser1275
Ser1309
Tyr960
Tyr1316
Tyr322
Tyr1165 Tyr1145
Tissue
Distribution
Present in most tissues Expressed in a variety of tissues A subset of neuronal tissues
Neuroblastomas
Neural crest derived sensory and sympathetic neurons
Subcellular
Localization
Plasma membrane Plasma membrane Plasma membrane
Binding Partner/
Associated Proteins
IRS1
IRS2
IRS3
IRS4
SHC
ADS
Sh2-B
Grb10
Grb7
CAP
p85 subunit of PI3K
SHPTP2 (Syp)
SOCS1-3
PC-1
SOCS1-3
14-3-3-ε/β/ζ
SHC
p85
IRS-1
IRS-2
IGF1R
JAK-1
PIK3R3
IGF-I
RACK1
PKCd
β-1 integrin
PKCμ
CSK
EHD1
NAG
ACP
AMP-PNP
Src
Not Known
Upstream
Activators
Insulin IGF-I Not Known
Downstream
Activation
IRS1-4
Shc
PTP1B
cbl
p62dok
IRS-1
IRS-2
Akt
and p42/44
MAPKs
PI3K
IRS-1
IRS-2
Ras
Activators Not Known Not Known Not Known
Substrates IRS 1-4
Gab-1
p62dok
Cbl
APS
Shc
Not Known Not Known
Selective
Inhibitors
Not Known Not Known Not Known
Non-Selective
Inhibitors
Hydroxy-2-naphthalenylmethylphosphonic acid
Quercetin (337951)
Staurosporine (S4400)
Not Known Not Known
Selective
Activators
Not Known Not Known Not Known
Physiological
Function
Insulin signaling Binds insulin-like growth factor I (IGF I) with a high affinity and IGF II with a lower affinity Embryonal development of dorsal root and trigeminal neurons
Development of sympathetic neurons
Male sexual differentiation
Disease
Relevance
Type A syndrome of insulin resistance
Leprechaunism
Rabson-Medenhall syndrome
Insulin resistance
Type-2 diabetes mellitus
Gastrointestinal stromal tumors
Crohn's disease
Primary breast cancer
Prostate cancer
Graves' disease
Pancreatic adenocarcinoma
Thyroid carcinomas
Colon cancer
Type-2 diabetes mellitus
Similar Products
References