Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
JAKs
The Janus kinases were initially identified as a novel subfamily of 120-140 kDa cytoplasmic tyrosine kinases that uniquely contained two, tandem domains with homology to tyrosine kinase catalytic domains. Initial insights into their roles in signal transduction came from genetic studies of interferon signaling and subsequently in biochemical studies of erythropoietin and growth hormone signaling. The most carboxyl-domain (JAK homology domain 1, JH1) has tyrosine kinase catalytic activity while the amino-terminal domain does not and is referred to as the pseudokinase domain (JH2). The amino-terminal half of the proteins contains the JH7-JH3 domains that are involved in receptor association, including a region (JH7-JH5) that contains a region with homology to a 300 amino acid protein-protein interaction domain referred to as the FERM domain (band four-point-one, ezrin, radixin, moesin domain).
The JAK family of kinases consists of four family members (JAK1, JAK2, JAK3, Tyk2) that each play unique roles in signal transduction. The major physiological roles of the JAKs are to initiate signal transduction through virtually all of the receptors of the cytokine receptor superfamily. The physiological roles of the JAKs have been most clearly illustrated through the derivation of mutant strains of mice lacking them. In particular, a deficiency of JAK2 causes loss of function of a variety of cytokine receptors including erythropoietin, thrombopoietin, growth hormone and prolactin, among a large number. Deficiency of JAK1 results in loss of function of most of the cytokine receptors that utilize receptors of the IL-6 cytokine receptor subfamily. Loss of Tyk2 results in some loss of interferon α/β signaling and a more profound loss of IL-12 signaling. As noted below a deficiency of JAK3 affects the IL-2 subfamily of cytokine receptors.
 The JAKs physically interact through their FERM domains with the cytoplasmic, membrane proximal regions of the receptors through a loosely conserved region referred to as the Box 1/Box 2 region. The specificity of these interactions is largely responsible for the specificity of physiological functions of the JAKs. For example, JAK3 is only known to associate with the common γ chain of the IL-2 receptor complex and consequently mutations/deletions of the JAK3 specifically eliminate signaling through the IL-2 subfamily of cytokine receptors that utilize this receptor chain. In general, single chain receptors require JAK2 or JAK1 for function while multi-chain receptors often require more than one JAK. For example, both JAK1 and JAK2 are required for interferon γ signal transduction while both JAK1 and JAK3 are required for the IL-2 receptor subfamily. The basis for the dependency on two JAKs in some receptor systems has not been established in biochemical detail. Receptor engagement results in changes in states of aggregation of the receptors, or conformation of their cytoplasmic domains, resulting in the trans-tyrosine phosphorylation of a regulatory site in the activation loop of the kinase domain causing mutual activation. Upon activation, the JAK kinases phosphorylate multiple sites on the kinase itself, on the receptor chains and substrates that are recruited to the receptor complex.
One of the common substrates of the JAK kinases in cytokine signaling are members of the signal transducers and activators of transcription (Stats) although a wide variety of signal transducing proteins have been shown to be phosphorylated by the JAKs in cytokine signaling. In humans, mutations of JAK3 are associated with genetically acquired severe combined immunodeficiency. Because of its importance in signaling by receptors controlling lymphoid cells, considerable interest has existed in defining specific inhibitors as immunoregulatory compounds. The activation of JAKs is frequently implicated in oncogenic transformation and human chromosomal translocations resulting in the activation of JAK2 have been identified. Although JAK activation has been reported in non-cytokine receptor superfamily receptor systems including both receptor tyrosine kinase and G-protein coupled receptor systems, the physiological relevance of this activation has not been definitively established through the use of the mutant strains of mice lacking JAKs.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Family Members JAK1 JAK2 JAK3 TYK2
Other Names Janus kinase 1 Janus kinase 2 Janus kinase 3 Tyrosine kinase 2
Molecular Weight 132 kDa
131 kDa
125 kDa
134 kDa
Structural Data 1142 aa 1132 aa 1124 aa 1182 aa
Isoforms Not Known Not Known JAK3M
JAK3B
JAK3S
Not Known
Species Vertebrate
Mammalian
Rat
Mouse metazoa
Plants
Fungi
Drosophila
Vertebrate
Mammalian
Rat
Mouse metazoa
Plants
Fungi
Drosophila
Vertebrate
Mammalian
Rat
Mouse metazoa
Plants
Fungi
Drosophila
Human
Mouse
Domain
Organization
7 JH homology domains
JH1 kinase domain
JH2 pseudokinase domain
7 JH homology domains
JH1 kinase domain
JH2 pseudokinase domain
7 JH homology domains
JH1 kinase domain
JH2 pseudokinase domain
7 JH homology domains
JH1 kinase domain
JH2 pseudokinase domain
Phosphorylation
Sites
Tyr1022
Tyr1023
Tyr1033
Tyr1007
Tyr1008
Tyr1033
Tyr980
Tyr981
Tyr1054
Tyr1055
Tissue
Distribution
Ubiquitous Ubiquitous Hematopoietic Ubiquitous
Subcellular
Localization
Cytoplasm Cytoplasm Cytoplasm Cytoplasm
Nucleus
Binding Partners/
Associated Proteins
Cytokine receptors
Growth factor receptors
Cytokine receptors
Growth factor receptors
Common γ chain Cytokine receptors
Growth factor receptors
Upstream
Activators
IFNα (I4401)
IFNγ (I3265)
IL-2 (I2644, I7908 (h))
IL-4 (I4269 (h))
IL-9
IL-13 (I1771 (h))
IL-15 (I8648)
IL-3 (I1646, I7389 (h))
IL-6 (I1395)
IL-11
CT-1
CNTF (C3710)
LIF (L5283)
OSM (O9635)
Prolactin (L4021)
IFNγ (I3265)
EPO (E5627)
IL-3 (I1646, I7389 (h))
IL-5 (I5273)
GM-CSF (G5035)
IL-6 (I1395)
IL-11 (I2406)
CT-1
CNTF (C3710)
IL-2 (I2644, I7908 (h))
IL-4 (I4269 (h))
IL-7 (I5896 (h))
IL-9
IL-15 (I8648)
IFNα (I4401)
IL-12 (I2276)
IL-13 (I1771 (h))
IL-6 (I1395)
IL-11 (I2406)
OSM (O9635)
CNTF (C3710)
LIF (L5283)
CT-1
Downstream
Activation
STATs/multiple pathways STATs/multiple pathways STATs/multiple pathways STATs/multiple pathways
Activators Not Known Not Known Not Known Not Known
Inhibitors Not Known AG 490 (T3434) CP-690550 (PZ0017)
JANEX-1
PNU15680
Not Known
Selective
Activators
Not Known Not Known Not Known Not Known
Physiological
Functions
Involved in interferon α/β/γ signaling pathway Signal transduction by multiple cytokines Signal transduction by IL-2 family cytokines Involved in initiation of type I IFN signaling
Disease
Relevance
Tumorigenesis
Leukemias
Myocardial ischemia
Diseases of abnormal erythropoiesis
Myeloproliferative disorders
Immunosuppressive diseases
Severe combined immunodeficiency
Lymphoproliferative disorders
Immune diseases
Similar Products
References