Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Biogenic Amine Transporters
Following vesicular release, the biogenic amine neurotransmitters, norepinephrine (A7257), dopamine (H8502) and serotonin (H9523), are removed from the extracellular space by selective and pharmacologically distinct transport proteins. These transporters, abbreviated NET, DAT and SERT, respectively, are of particular clinical interest because they are the molecular targets for many antidepressants as well as drugs of abuse, such as cocaine and the amphetamines. The molecular cloning of NET, DAT and SERT revealed that these transporters arise from single genes with homology to members of a larger sodium-dependent transporter gene family. Structural features common to these transporters are a 12 transmembrane-spanning domain structure with intracellular amino- and carboxy-tails that has been largely confirmed by site-specific labeling techniques. Multiple potential phosphorylation sites exist on intracellular domains, supporting evidence of kinase-mediated regulation of these transporters. In addition, amphetamines and other substrates for biogenic amine transporters appear capable of regulating transporter cell surface expression perhaps via activity-dependent processes, thereby establishing both endogenous and pharmacological regulatory mechanisms. Further regulation of biogenic amine transporter function may come from the formation of homo-oligomers or the associations with other membrane proteins to form heteroligomers. As our understanding of these protein-protein interactions evolves, significant efforts will be focused on elucidating the structural requirements for these interactions and how drugs may influence associations to regulate function.
The availability of cDNAs for NET, DAT and SERT has permitted detailed pharmacological study of each protein in heterologous expression systems. Interestingly, both NET and DAT transport norepinephrine and dopamine with relatively high affinity; thus, distinction between NET and DAT relies upon sensitivity to transporter antagonists and anatomical localization. Whereas NET is inhibited by the tricyclic antidepressants, including desipramine and nortriptyline as well as more selective drugs such as nisoxetine, DAT is relatively insensitive to these agents but is potently inhibited by GBR 12909 and GBR 12935, compounds that demonstrate lower affinity for NET. SERT shows the greatest substrate selectivity with >1000-fold higher affinity for serotonin as compared to the other biogenic amines. SERT is most potently inhibited by the highly prescribed class of selective serotonin reuptake inhibitors (SSRIs) that include paroxetine, fluoxetine, and sertraline. All of the biogenic amine transporters are inhibited with approximately equal potency by cocaine, suggesting commonality among the transporters with regards to cocaine recognition. With regards to general structure-activity relationships for biogenic amine transporter ligands, studies have consistently demonstrated the importance of substrate and antagonist amine groups for high-affinity interactions. However, a novel class of high-affinity transporter ligands has been described that lack amine groups, challenging the requirement for amine-containing structures and suggesting new directions in the design and synthesis of biogenic amine transporter ligands.
By comparison to the plasma membrane monoamine transporters, the vesicular monoamine transporters (VMATs), while retaining a predicted 12 transmembrane-spanning domain structure, are members of a separate proton-dependent gene family of transporters with distinct pharmacological sensitivity. This family of vesicular neurotransmitter transporters also contains the vesicular acetylcholine transporter. The two VMATs, VMAT-1 and VMAT-2, have been cloned and demonstrate broad substrate recognition for monoamine neurotransmitters. The major differences between VMAT-1 and VMAT-2 are in their tissue localization, with VMAT-1 primarily found in endocrine cells and VMAT-2 localized to neuronal tissues. Pharmacologically, VMAT-2 is inhibited by reserpine (R0875) and tetrabenazine (T2952), whereas VMAT-1 is relatively insensitive to inhibition by tetrabenazine. In general, these vesicular transporters are thought to play a major role in packaging neurotransmitters into distinct secretory vesicles in preparation for subsequent exocytotic release, thus controlling quantal size of each release event. The important role that these transporters play in neurotransmission has led to their implication in psychiatric and neurodegenerative disorders and fueled interest in developing selective pharmacological agents targeted to these transport proteins.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Currently Accepted Name Dopamine transporter
(DAT)
Norepinephrine transporter
(NET)
Serotonin transporter
(SERT)
 (S1943)
Vesicular monoamine transporters
(VMATs)
Structural Information 620 aa (human) 617 aa (human) 630 aa (human) 525 aa (human VMAT-1)
514 aa (human VMAT-2)
Uptake Inhibitors GBR-12909a (D052)
Nisoxetinea (N151)
GBR-12935a (G9659)
Indatraline (Lu-19-005) (I119)
Bupropion (B102)
Amfonelic acid
BTCP (B138)
Mazindol (M2017)
Nomifensine (N1530)
β-CFT (WIN 35,428) (C124)
β-CPT (WIN 35,065-2) (C156)
β-CIT (RTI-55)
GYKI 52895 (G120)
4',4''-Difluoro-3a-diphenyl-methoxytropane
4'-Chloro-3a-diphenylmethoxy-tropane
Cocaine (C5776)
Citaloprama (C7861)
Tomoxetinea (T7947)
Desipramine (D3900)
Nortriptyline (N7261)
Protriptyline (P8813)
Imipramine (I7379)
Xylamine
Nomifensine (N1530)
Mazindol (M2017)
Amoxapine (A129)
Indatraline (Lu-19-005) (I119)
Reboxetine (R6527)
Maprotinine (M9651)
Duloxetine (SML0474)
Venlafaxine (V7264)
Cocaine (C5776)
Reserpine (R0875)
6-Nitroquipazinea (Q109)
Paroxetinea (P1372)
Sertralinea (S6319)
Fluoxetinea (F132)
Clomipramine (C7291)
Imipramine (I7379)
Alaproclate (A164)
Trazodone (T6154)
Zimelidine (Z101)
Indatraline (Lu-19-005) (I119)
Fluvoxamine (F2802)
Venlafaxine (V7264)
β-CIT (RTI-55)
Nefazodone (N5536)
LY-367,265 (L2411)
Duloxetine (SML0474)
Cocaine (C5776)
Tetrahydrobenazine
Tissue Expression Brain Brain
Adrenal gland
Placenta
Brain
Platelet
GI tract
Placenta
Brain
Endocrine cells
Adrenal gland
GI tract
mast cells  
Physiological Function These transporters regulate the temporal and spatial actions of neurotransmitters by removing them from the extracellular space VMATs package neurotransmitters into secretory vesicles in preparation for exocytotic release, thus controlling quantal size of each release event
Disease Relevance Parkinson's disease
Drug abuse
Schizophrenia
Affective disorders
Attention-deficit/hyperactivity disorder (ADHD)
Drug abuse
Neuropathic pain
Affective disorders
Anxiety disorders
Neuropathic pain
Drug abuse
Hypertension
Parkinson's disease
Drug abuse  
Footnote
a) Best characterized, selective inhibitors for each transporter.
Abbreviations
BTCP: N-[1-(1-Benzo[β]thien-2-ylcyclohexyl)]piperidine
β-CFT: 2β-Carbomethoxy-3β-(4-fluorophenyl)tropane
β-CIT: 2β-Carbomethoxy-3β-(4-iodophenyl)tropane
β-CPT: 2β-Carbomethoxy-3β-phenyltropane
GBR-12909: 1-[2-[bis(4-Fluorophenyl)methoxy]ethyl]-4-[3-phenylpropyl]piperazine
GBR-12935: 1-(2-[Diphenylmethoxy]ethyl)-4-[3-phenylpropyl]-piperazine
GYKI 52895: 1-(4-Aminophenyl)-4-methyl-7,8-methylenedioxy-3,4-dihydro-5H-2,3-benzodiazepine
LY-367,265: 1-[2-[4-(6-fluoro-1H-indol-3-yl)-3,6-dihydro-1(2H)-pyridinyl]ethyl]-5,6-dihydro-1H,4H-[1,2,5]thiadiazolo[4.3.2-ij]quinoline-2,2-dioxide
Similar Products
References