Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Anti-HA-Fluorescein Troubleshooting
Product No. 11666878001
Troubleshooting
Signal week or missing; too few cells stained
  • Protein is not expressed in cells. → Prepare a cell extract and use western blot analysis to verify that tagged protein is expressed.
  • Too few cells expressing protein → Use more cells in sample. Try a different transient transfection procedure or use a stably transfected cell line.
  • Poor cell permeability → Increase time of incubation with PBS/Triton.
  • Sample fixation before staining destroys epitope → Use a different sample fixation method.
  • Tag-specific antibody too dilute → Run a titration curve (identical samples, varying amounts of tag-specific antibody, optimal amounts of secondary antibody) to determine the optimal dilution of tag-specific antibody.
High background
  • Antibody too concentrated → Run a titration curve (identical samples, varying amounts of tag-specific antibody, optimal amounts of secondary antibody) to determine the optimal dilution of tag-specific antibody.
  • BSA in Blocking Solution contains IgG → Use only high purity (IgG-free) BSA.