Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
cOmplete™ Lysis-M EDTA-free Troubleshooting
Product No. 04719964001
Troubleshooting
Protein concentration in lysate
The low protein concentration in cell lysates may be an indication that the volume of lysis buffer is not sufficient to completely lyse the cells. In this case, do not decrease the volume of Complete inhibitor. For best results, use the volumes indicated in the package insert. For a 6-well plate, use 200-400 μl per well. If the protein concentration in the lysate is still low, increase the incubation time and shake the mixture more vigorously.
When there are low cell numbers in your primary cultures (because confluence is not reached), this can also be the reason for low protein content. to increase protein yield, scrape the cells from the plate and collect as many cells as possible using centrifugation. Remove as much of the supernatant as possible, and then find the optimum volume of Complete inhibitor by lysing the cell pellet in 50, 100, or 150 μl volumes.
Materials
COMPLETE is a trademark of Roche
Related Links