Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Sterile Millex® Filters for Cell Culture
Millex syringe filters are ideal for sterilizing 1 mL – 200 mL of cell culture reagents, such as cell culture media, antibiotics, supplements, sera, viruses for infection, and other biological solutions. Many sterile Millex filters are CE-marked and/or are approved as medical devices, meaning that they can be used to filter pharmaceutical solutions before they are introduced into human patients.
Because of the quality of their design, manufacturing, and quality control, sterile Millex filters are known for superior performance compared to syringe filters from other suppliers:
  • Less clogging, faster flow
  • Minimal leaching of impurities into filtered solutions
  • Minimal nonspsecific binding of desirable biomolecules, such as high value antibiotics, growth factors, and serum proteins
Sterile Millex syringe filters are now available here on sigmaaldrich.com in a variety of membrane types, diameters, and pore sizes. We are continuously launching more and more Millipore products on this site to enhance your purchasing experience.
Product No. Membrane Pore Size (µm) Diameter (mm) Process Volume (hold-up) Housing & Sterilization Method CE Marked/ Medical Device Pack Size
SLLG025SS PTFE, Hydrophilic Broad chemical compatibility 0.2 µm Sterile Filtration of DMSO 25 mm 100 mL (<100 µL) HDPE, EO   50 units/pk
SLGV013SL Durapore® (PVDF) Membrane Lowest binding membrane for protein-rich solutions 0.22 µm Sterile Filtration 13 mm 10 mL (< 50 µL) HDPE, EO CE 100 units/pk
SLMPL25SS Millipore Express® PLUS (PES) Membrane Fast flow and low binding for cell culture media preparation 0.22 µm Sterile Filtration 25 mm 100 mL (<100 µL) PVC, EO CE 50 units/pk
SLMP025SS Millipore Express PLUS (PES) Membrane Fast flow and low binding for cell culture media preparation 0.22 µm Sterile Filtration 25 mm 100 mL (<100 µL) PVC, EO M 50 units/pk
SLGSV255F MF-Millipore™ (MCE) Membrane Most referenced general purpose membrane 0.22 µm Sterile Filtration 25 mm 100 mL (<100 µL) PVC, vented inlet, EO   50 units/pk
SLGL0250S MF-Millipore (MCE) Membrane Most referenced general purpose membrane 0.22 µm Sterile Filtration 25 mm 100 mL (<100 µL) PVC, vented inlet, EO   50 units/pk
SLGV004SL Durapore (PVDF) Membrane Lowest binding membrane for protein-rich solutions 0.22 µm Sterile Filtration 4 mm 1 mL (<10 µL) HDPE, EO   100 units/pk
SLHV013SL Durapore (PVDF) Membrane Lowest binding membrane for protein-rich solutions 0.45 µm Clarification of sterile solutions 13 mm 10 mL (< 50 µL) HDPE, EO CE 100 units/pk
SLHV004SL Durapore (PVDF) Membrane Lowest binding membrane for protein-rich solutions 0.45 µm Clarification of sterile solutions 4 mm 1 mL (<10 µL) HDPE, EO   100 units/pk
SLSV025LS Durapore (PVDF) Membrane Lowest binding membrane for protein-rich solutions 5.0 µm Clarification of sterile solutions 25 mm 100 mL (<100 µL) PVC, EO   50 units/pk
SLAAV255F MF-Millipore (MCE) Membrane Most referenced general purpose membrane 8.0 µm Clarification of sterile solutions 25 mm 100 mL (<100 µL) PVC, vented inlet, EO   50 units/pk
Product No. Membrane Pore Size (µm) Diameter (mm) Housing & Sterilization Method Inlet Connection Outlet Connection Pack Size
SLFG025LS Fluoropore™ (PTFE) Membrane Hydrophobic chemistry for gas filtration 0.20 µm 25 mm PVC, EO FLL MLS 50 units/pk
SLGVS25PS Durapore (PVDF) Membrane Super hydrophobic membrane for transducer protection 0.22 µm Sterile Filtration 25 mm PVC, EO FLL MLS 50 units/pk
SLGVS25US Durapore (PVDF) Membrane Super hydrophobic membrane for transducer protection 0.22 µm Sterile Filtration 25 mm PVC, EO FLL MLL 50 units/pk
SLGVS25XS Durapore (PVDF) Membrane Super hydrophobic membrane for transducer protection 0.22 µm Sterile Filtration 25 mm PVC, EO FLL MLL 50 units/pk
SLGVS25LS Durapore (PVDF) Membrane Super hydrophobic membrane for transducer protection 0.22 µm Sterile Filtration 25 mm PVC, EO FLL Spike 50 units/pk
* Male Luer-Lok®