Elix® Advantage for Water Purification
The most advanced pure water system producing Elix® water quality
Elix® Advantage has been designed to deliver the highest pure water quality for your regular laboratory applications and instrument feed. With over 20 years of proven success, Elix® is suited to every user’s needs in today’s demanding laboratory environment.
Elix® Advantage produces pure, Type 2 water from potable water. It features two separate and distinct components:
  • The production unit – compact, it can be placed wherever it best suits you.
  • The E-POD® remote dispenser, Elix®-Point-Of-Delivery unit – convenient and always within reach, it brings pure, bacteria-free water at 2 L/min right where you need it.
The system has the capacity to produce up to 300 L/day
Features and Benefits
  • The Elix® Advantage system is the first choice of scientists around the world for pure water in both validated and non-validated environments. Elix® water quality is ideal for the preparation of reagents and buffers, for microbiology media or histology. It is also the best feed for all Milli-Q® ultrapure systems, as well as instruments such as glassware washers, autoclaves, hydrogen generators.
  • Thanks to the combination of technologies inside, Elix® water purification allow to meet the highest users' requirements in terms of water purity, ergonomy thanks to the E-POD® remote dispenser, validation and traceability.
Water Applications
Pure Water Applications
  • Biochemistry automation
  • Clinical analyzers
  • Dissolution testing
  • Fog generation
  • General laboratory applications
  • Glassware rinsing
  • Microbiological culture media
  • Production
  • Stability chambers
  • Weathering Test Chambers
Water Specifications
Elix® Advantage pure water (Type 2)
Resistivity at 25 °C*
Typically 10-15 MΩ•cm
TOC
< 30 ppb
From an E-POD® dispenser with final filter, the following water quality specifications are achieved:
Particulates (size > 0.22 µm)**
< 1 particulate/mL
Bacteria** < 0.1 CFU/mL
Pyrogens (endotoxins)*** < 0.001 EU/mL
RNases*** < 0.01 ng/mL
Dnases*** < 4 pg/µL
Flow rate Up to 2 L/min
* Resistivity can also be displayed non-temperature-compensated as required by USP
** With Millipak® final filter
*** With Biopak® Polisher