POD-Paks: Water Quality to Match All Your Application Needs
Do you use Milli-Q® water and perform a variety of applications in your lab?
You'll find that our range of application POD-Paks is the perfect solution. Each of the POD-Pak polishers targets specific types of contaminants and removes them right at the Q-POD® dispenser.
The POD-Pak range includes:
POD-pak range: Millipak & sterile Millipak Gold, Biopak, VOC-Pak, EDS-Pak, LC-Pak
POD-Pak Range: Millipak® & Sterile Millipak® Gold, Biopak®, VOC-Pak®, EDS-Pak®, LC-Pak®
Once the appropriate Application-Pak is installed, your Milli-Q® IQ 7000 will provide water suitable for most of the analytical techniques used in the laboratory, including:
  • HPLC
  • LC-MS
  • UPLC
  • MALDI-ToF-MS
  • IC
  • ICP
  • GC
  • AA
  • CE
  • Particle analysis
  • Surface tension
  • Spectrophotometry
In addition, specific Application-Paks are available for use with life sciences applications, such as:
  • Cell Culture
  • Western blotting
  • Dissolution testing
  • Spectroscopy
  • Immunoassays
  • PCR
  • Sequencing
  • Proteomics
  • Genomics
  • Microbiology
The new application POD-Paks features include:
  • An e-Sure tag, for indispensable traceability.
    Consumable data is automatically recorded in the system's memory, allowing consumable status to be monitored on the Q-POD® screen at any time. Data includes installation date, pack ID, and events throughout the consumable's lifetime.
  • An ergonomic pack locker — for easy "snap-into-place" installation
If you need several types of ultrapure water, simply connect up to four Q-POD® dispensers to your system, each with a different Application-Pak.
Millipak interface
Millipak interface