Immunochemical Analysis with Cell Microarrays | Biowire Spring 2011
Biowire Spring 2011 — Cell Lines — Models of Disease
Subscribe to Biowire and eBiowire
Download Biowire Spring 2011 (7.0 Mb PDF)
Biowire Spring 2011 Home
Immunohistochemistry on Tissue and Cell Microarrays
Immunohistochemistry (IHC) is a widely used method for determining the level and distribution of protein expression in tissues and cells. With the development of the tissue microarray (TMA) technology, IHC can now be used for the simultaneous analysis of protein expression in a multitude of tissue samples. Within the ongoing Human Protein Atlas (HPA) project (proteinatlas.org)1,2,3, the human proteome is systematically analyzed in 48 normal human tissues and 20 different cancer types using TMAs and automated IH C in a high-throughput setting.
The ability to embed cultured cells in paraffin enabled the construction of cell microarrays (CMAs). As a complement to the TMA, a CMA, including 47 human cell lines and 12 leukemia cell samples, was designed within the HPA project4. An objective of this design is to facilitate studies of normal hematopoiesis and differentiation as well as tumor progression. Commonly used cell lines were selected to represent the major forms of solid tumors and hematological malignancies. Cell types not represented in the HPA tissue microarrays were also included. In addition to providing more comprehensive protein expression profiles, CMAs also render many uncontrolled variables equal across the cells, thus improving the possibility for relative protein quantification.
back to top Back to Top
The Human Protein Atlas
Establishment of the HPA project permits a systematic genome-based exploration of the human proteome using antibodybased proteomics. This is accomplished by combining high-throughput generation of Prestige Antibodies with protein profiling in a multitude of human tissues and cells. In November 2010, Prestige Antibodies were used to analyze protein expression of more than 10,000 human genes. Each year, protein expression and localization data for approximately 2,500 new proteins are added to the portal. By 2015, a first draft of the spatial expression profile of the human proteome will be completed.
The Human Protein Atlas is a public web portal managed by an academic project that aims to map the human proteome in a period of 10 years. Today, more than 700 IHC, WB, and IF images are presented for each of 13,200 antibodies against human targets, covering 50% of the human proteome.
Antibodies developed and characterized within the Human Protein Atlas project are available to the scientific community as Prestige Antibodies. Prestige Antibodies are co-exclusively distributed by Sigma-Aldrich® and Atlas Antibodies in Europe, and exclusively distributed by Sigma-Aldrich outside of Europe.
back to top Back to Top
Cell Microarrays
Cell microarrays are constructed to allow simultaneous analysis of protein expression in a multitude of cell samples. To enable the assembly of in vitro cultured cell lines and primary cells in a microarray format, the cells are first homogeneously distributed in an agarose gel matrix. After polymerization of the agarose, the cell-agarose gel is fixed in 4% formaldehyde and histoprocessed using a vacuum infiltration processor. With this protocol, artificial tissue blocks can be used in a similar manner as paraffin-embedded tissues for cell microarray construction.
The actual CMA is subsequently produced by extracting 0.6 mm cylinders from the cell blocks with a sharp punch and placing them in a recipient block with a core-tocore distance of 1.3 mm in all directions. An automatic TMA arrayer is used for the CMA construction. From one array block, approximately 250 sections can be cut and prepared for immunochemical analysis.
All Prestige Antibodies® are analyzed on sections from the CMAs. The resulting protein expression profiles and all immunostaining images showing cellular distribution (Figure 1) are shown on the Human Protein Atlas portal (proteinatlas.org).
Figure 1. Immunochemical staining showing subcellular localization patterns. 
Figure 1. Immunochemical staining showing subcellular localization patterns.
The specific binding of an antibody to its corresponding antigen results in a brown-black staining by a horseradish peroxidase reaction. Blue color of nuclei corresponds to unspecific counter-staining with hematoxylin. A) Anti-Emerin (HPA000609) stains nuclear membrane in cell line RT-4; B) Anti-CSTF2 (HPA00427) stains nucleus in cell line PC-3; C) Anti-VSX2 (HPA03436) stains nucleoli in cell line BEWO; and D) Anti-NF2 (HPA003097) stains plasma membrane in cell line U-2 OS.
back to top Back to Top
Image Analysis
The purpose of using CMAs is to gain protein localization information on cellular and subcellular levels, and to achieve relative quantification results. In order to generate non-subjective quantitative data from immunohistochemistry, analysis of immunoreactivity in cells and cell lines in the CMA is performed using automated image analysis5.
Immunostained CMA slides are scanned to generate high-resolution digital images. Automated scanning is performed using 40x magnification. Output parameters from the software are always displayed in conjunction with the annotated images and include: number of objects defined as cells in the image; staining intensity (negative, weak, moderate, and strong); and fraction (%) of positive cells. Two overlay images with additional numerical information are presented to facilitate interpretation (Figure 2).
Figure 2. Automated Image Analysis of HPA005785 on the Human Protein Atlas 
Figure 2. Automated Image Analysis of HPA005785 on the Human Protein Atlas.
A) Immunochemical staining of Anti-CD44 shows positivity of plasma membrane. B) Object-based view representing fraction (%) of immunostained cells. The color code for each cell represents a range of immunoreactivity: blue (negative/ very weak), yellow (weak/moderate), orange (moderate/strong), and red (strong) cells. This classification is based on areas of different intensities within each object (cell). C) Area-based view representing immunostained areas (%) within cells. The color code represents a range of immunoreactivity: yellow (weak/moderate), green (moderate/strong), and red (strong). Negative/very weak areas are transparent. The annotated intensity score presented on the HPA is deduced from data generated from both the object-based and the area-based analysis.
back to top Back to Top
Cell Expression Diagram
A cell expression diagram is presented for each Prestige Antibody on the Human Protein Atlas (Figure 3). This diagram represents a visualization of normalized protein expression values across the different cells relative to the cell type with the highest level of expression, e.g., PBMC (M36) in Figure 3. Cells are grouped according to cellular origin to facilitate functional analysis and identification of proteins with a cell-type-specific expression pattern. Since protein expression correlates with cellular size, the diagram is based on expression levels normalized against the total protein content for each particular cell type (expression values for all analyzed antibodies added together in each cell type)5.
Figure 3. Cell Expression Diagram displayed on the Human Protein Atlas visualizing the expression level for the Emerin protein by HPA000609 in the analyzed cell lines and cell samples as estimated using automated image analysis. 
Figure 3. Cell Expression Diagram displayed on the Human Protein Atlas visualizing the expression level for the Emerin protein by HPA000609 in the analyzed cell lines and cell samples as estimated using automated image analysis.
back to top Back to Top
Summary
CMAs enable simultaneous immunochemical analysis of protein expression using Prestige Antibodies in 59 cell lines and primary cell samples.
Protein localization information on cellular and subcellular levels can be achieved by immunochemical analysis on CMAs using Prestige Antibodies.
118 high-resolution immunochemical images from 12 clinical leukemia cell samples and 47 human cell lines are presented for each Prestige Antibody on the Human Protein Atlas portal.
Automated image analysis objectively scores protein expression.
Standardized protocols, the use of CMAs, and normalization in reference to total protein expression enable extrapolation of quantitative immunochemical data.
back to top Back to Top
References
  1. Uhlén M, et al. A Human Protein Atlas for Normal and Cancer Tissues Based on Antibody Proteomics. Mol Cell Proteomics. 2005;4(12):1920–32.
  2. Berglund L, et al. A gene-centric human protein atlas for expression profiles based on antibodies. Mol Cell Proteomics. 2008;7:2019–27.
  3. Pontén F, et al. The Human Protein Atlas - a tool for pathology. J Pathology. 2008;216(4):387–93.
  4. Andersson AC, et al. Analysis of protein expression in cell microarrays: a tool for antibody-based proteomics. J Histochem Cytochem. 2006;54(12):1413–23.
  5. Lundberg E, et al. The correlation between cellular size and protein expression levels — Normalization for global protein profiling. J Proteomics. 2008;16(5):493–502.
back to Biowire main page
back to top Back to Top
Related Links