Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Air-Stable Precatalysts for Amination
By: Josephine Nakhla, Chemfiles Volume 10 Article 3
C–N bond forming cross-couplings typically require a palladium source along with the associated ligands. Most Pd(0) sources are not air-stable, while the commonly employed air-stable Pd(0) source, Pd2(dba)3 contains associated ligands which could impede the reaction. Stable Pd(II) precursors require reduction under the reaction conditions. In either case, a ligand must be added to the reaction in order to lead to the active Pd-species. Buchwald and coworkers recently reported the use of highly-active yet air- and moisture-stable precatalysts, which, under the standard reaction conditions, form the active monoligated Pd-species. These precatalysts (704954, 704946, 707589, 708739) are exceptionally efficient even under challenging conditions, such as coupling electron-poor anilines with deactivated aryl chlorides. The catalyst precursors also offer other advantages including low catalyst loadings and short reaction times.
Scheme 2. Buchwald precatalysts in N-arylations of electron-poor amines with electron-rich aryl chlorides
Scheme 2. Buchwald precatalysts in N-arylations of electron-poor amines with electron-rich aryl chlorides (704954) (707589)
Scheme 3. Buchwald precatalysts in N-arylations with low catalyst loadings
Scheme 3. Buchwald precatalysts in N-arylations with low catalyst loadings (704954)
back to top
Materials
References
  1. Biscoe, M. R. et al. J. Am. Chem. Soc. 2008, 130, 6686.
For more information on our Buchwald catalyst and ligand portfolio, visit aldrich.com/buchwald