Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Alkyne Hydration
By: Josephine Nakhla, ChemFiles Volume 10 Article 4
The hydration of alkynes has been extensively studied for more than 100 years. This reaction allows access to various carbonyl derivatives from alkyne precursors. Nolan and coworkers reported alkyne hydration using a gold catalyst. Nolan, a pioneer in the use of N-heterocyclic carbenes (NHCs) as ligands in various catalytic transformations with diff erent metals, developed conditions employing a gold-NHC complex and silver hexafl uoroantimonate for the hydration of alkynes (Scheme 1). It is important to note that an acid is not needed for this transformation and the reaction was feasible at low catalyst loadings (10 ppm).
Scheme 1 Hydration of alkynes using gold-NHC complex
back to top Back to Top
References
  1. Marion, N.; Ramón, R. S.; Nolan, S. P. J. Am. Chem. Soc. 2009, 131, 448.
back to top Back to Top
Related Links