β-Amino Acids and Homologs
By: Matthias Junkers, Aldrich ChemFiles 2008, 8.7, 11.
Aldrich ChemFiles 2008, 8.7, 11.
Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) might be the most thoroughly investigated peptidomimetics. Besides being remarkably stable to metabolism, exhibiting slow microbial degradation, and inherently stable to proteases and peptidases, they fold into well-ordered secondary structures consisting of helices, turns, and sheets. In this respect, the most intriguing effects have been observed when β2-amino acids are present in the β-peptide backbone.1 A whole new “world” has emerged from the design of fascinating new peptidic macromolecules from β- and γ-homologated proteinogenic amino acids and other components.2 Sigma-Aldrich® is proud to be a leading supplier of β-amino acids. This ChemFiles presents a comprehensive list of available products.
back to top Back to Top
Materials
References
  1. Lelais, G.; Seebach, D. Biopolymers 2004, 76, 206.
  2. Seebach, D.; Beck, A.K.; Bierbaum, D.J. Chem. & Biodiv. 2004, 1, 1111.
back to top Back to Top
Related Links