Asymmetric 1,3-dipolar Cycloaddition Using Fesulphos
By: William Sommer, Aldrich ChemFiles 2008, 8.6, 14.
Aldrich ChemFiles 2008, 8.6, 14.
The asymmetric 1,3-dipolar cycloaddition reaction is of the utmost importance for the enantioselective synthesis of five-membered heterocycles. Cabrera et al. introduced a new family of ligands consisting of a planar chiral P,S-ligand, named Fesulphos, for the 1,3-dipolar cycloaddition of azomethine halides.1 The catalytic reaction is carried out with the Fesulphos ligand, a copper salt, and triethylamine in methylene chloride. This new catalytic system demonstrated complete enantiocontrol (ee >99%) with conversion up to 97% (Scheme 1).
Scheme 1.