Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Brominated Polystyrenes and Bromoalkylated Resins
Chemfiles Volume 3 Article 4
Brominated polystyrene is a core matrix for the preparation of various functionalized polystyrene resins for SPPS and SPOS. The brominated polymer can be derivatized by Suzuki coupling reactions(1) or lithiation followed by reaction with electrophiles.(2) Acylation of bromoalkylated supports may be accomplished using the carboxylic acid cesium salts in the presence of KI.(3)
back to top 
Materials
References
  1. Woolard, F. X. et al., J. Org. Chem., 1987, 62, 6102.
  2. Farrall, M. J. et al., J. Org. Chem., 1976, 41, 3877.
  3. Gisin, B.F., Anal. Chim. Acta 1972, 58, 248.