Cationic Palladium Complex, [Pd(dppp) (PhCN)2](BF4)2 in Hetero Diels-Alder Reaction
By: Josephine Nakhla, Chemfiles volume 10 Article 3
Cationic palladium(II) complexes are utilized in a variety of reactions. [Pd(dppp)(PhCN)2](BF4)2 (696617) was shown to catalyze the hetero Diels-Alder reaction of dienes with aldehydes. The reaction yields substituted 5,6-dihydro-2H-pyrans without the use of Lewis acids (Scheme 1). The reaction is believed to proceed through a stepwise mechanism.
Scheme 1. Cationic palladium complex in hetero Diels-Alder reaction
Scheme 1. Cationic palladium complex in hetero Diels-Alder reaction (696617)
back to top
Materials
References
  1. Oi, S.; Kashlwagi, K.; Terada, E.; Ohuehi, K.; lnoue, Y. Tetrahedron Lett. 1996, 37, 6351.
  2. Davies, J. A.; Hartley, F. R.; Murray, S. G. J. Chem. Soc. Dalton Trans. 1980, 2246.
For more information on Aldrich Chemistry's catalyst portfolio, please visit aldrich.com/catalysis