Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
CBS Catalysts
Aldrich ChemFiles 2005, 5.4, 2.
Aldrich ChemFiles 2005, 5.4, 2.
2-Methyl-CBS-oxazaborolidine and o-tolyl-CBS-oxazaborolidine
Since 1987, the series of chiral oxazoborolidines known as CBS catalysts (after Corey, Bakshi, and Shibata) have been used for catalytic reduction of prochiral ketones,1 imines,2 and oximes3 to produce chiral alcohols, amines, and amino alcohols, respectively, in excellent yields and ee’s. Sigma-Aldrich is pleased to offer both enantiomers of 2-methyl-CBS-oxazaborolidine as dry reagents, as well as 1 M solutions in toluene.
We are also pleased to offer o-tolyl-CBS-oxazaborolidine as a 0.5 M solution in toluene for your research needs. When protonated with trifluoromethanesulfonimide, these chiral oxazaborolidines generate chiral Lewis acids, which have demonstrated great utility in the enantioselective Diels–Alder reaction (Scheme 1)4.
Scheme 1
back to top 
Materials
References
  1. (a) Corey, E. J. et al. J. Am. Chem. Soc. 1987, 109, 5551; (b) Corey, E. J. et al. J. Am. Chem. Soc.1987, 109, 7925.
  2. (a) Kirton, E. H. M. et al. Tetrahedron Lett. 2004, 45, 853; (b) Cho, B. T.; Chun, Y. S. Tetrahedron: Asymmetry 1992, 3, 337; (c) Cho, B. T.; Chun, Y. S. J. Chem. Soc., Perkin Trans. 1 1990, 3200.
  3. Tillyer, R. D. et al. Tetrahedron Lett. 1995, 36, 4337.
  4. Ryu, D. H.; Corey, E. J. J. Am. Chem. Soc. 2003, 125, 6388.