Chiral Phospholane Ligands
Aldrich ChemFiles 2007, 7.3, 15.
Aldrich ChemFiles 2007, 7.3, 15.
Chiral phospholane ligands have been used extensively in transition metal-catalyzed asymmetric hydrogenations and other novel asymmetric reactions including [4+1] cycloadditions, imine alkylations, and allylborations.
Available in either enantiomeric form with Me, Et, and i-Pr substituents. For a detailed product listing, visit sigma-aldrich.com/phospholane.