Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Cyclic Aryl-Substitued Amino Acid Derivatives
ChemFiles Volume 1 Article 1
Aromatic a-substituted proline analogs and g-substituted pyroglutamates are valuable building blocks in both medicinal and peptide chemistry. Through Suzuki coupling technology, they can be used as starting materials in the conversion of aryl bromide derivatives into biaryl compounds. These products have also found uses in peptide chemistry for the introduction of diversity and structural constraints in peptidomimetics.