CYPHOS® Phosphonium Ionic Liquids
Aldrich ChemFiles 2005, 5.6, 15.
Aldrich ChemFiles 2005, 5.6, 15.
The phosphonium cation, with the generic formula [PR3R’]+ and with a judicious selection of the appropriate anion, forms many phosphonium salts that are liquid at room temperature and many have melting points below 100 °C.
Phosphonium-based Ionic Liquids offer the following advantages:
  • Phosphonium salts are thermally more stable than the corresponding ammonium salts and imidazolium salts. This is very important for processes which operate at temperatures higher than 100 °C (e.g., removal of reaction products by distillation). Furthermore, prevention of contaminations by decomposition products is a prerequisite for multiple recycle use of solvents.
  • Phosphonium cations lack acidic protons. Thus, they are stable under basic conditions, which can be problematic for several imidazolium-based Ionic Liquids (carbene formation).
  • Phosphonium salts are, in general, less dense than water, which might be beneficial in product work-up steps.
Sigma-Aldrich offers, in collaboration with Cytec Industries, Inc., a new class of Ionic Liquids based on phosphonium cations.
back to top
Materials
References
  1. Bradaric, C. J.; Downard, A.; Kennedy, C.; Robertson A. J.; Zhou, Y. Green Chem. 2003, 5, 143