DuPhos and BPE Phospholane Ligands
Aldrich ChemFiles 2006, 6.8, 3.
Aldrich ChemFiles 2006, 6.8, 3.
Introduction
Asymmetric hydrogenation reactions represent the ideal process for the commercial manufacture of single-enantiomer compounds, because of the ease by which these robust procedures can be scaled up and because of the low levels of byproducts generated in these asymmetric processes. The most effective hydrogenation systems rely on modifications of the electronic and steric properties of the ligands. Burk and co-workers succeeded in developing a highly effective chiral phospholane class of ligands called DuPhos and BPE that contain 2,5-disubstituted groups allowing for systematic variation of the steric environment around the metal.1 Sigma-Aldrich, in collaboration with Kanata Chemical Technologies, is pleased to now offer a diverse array of DuPhos and BPE phospholane ligands that can be ligated to metal complexes to afford highly active catalysts for asymmetric hydrogenation and other innovative transformations.2
The large-scale capacity of these robust catalysts is observed in the efficiency (substrate-to-catalyst (S/C) ratios up to 50,000) and the high activities (TOF > 5000 h-1) in a myriad of enamide and ketone reductions. Under optimized conditions, a (R,R)-Me-BPE-Rh complex reduced N-acetyl a-arylenamides in >95% ee to yield valuable a-1-arylethylamines (Scheme 1).3 It should be noted that Me-DuPhos-Rh complexes were equally effective in asymmetric reductions of prochiral enamides. The general utility of these phospholane ligands is illustrated in the incredible diversity-oriented production of a vast array of chiral compounds (Scheme 2).
Scheme 1
Scheme 2
back to top
Advantages of the DuPhos and BPE Ligands
  • Superior enantiocontrol in a vast array of catalytic transformations
  • High activities at low catalyst loadings
  • Exceptional chemoselectivities for specific reaction paradigms
  • Asymmetric hydrogenations of numerous unsaturated substrates
  • First-to-market exclusivity for selected portfolio ligands
  • Screening kit to facilitate tuning selectivities (Fall 2006)
back to top
Representative Applications
The reactivity profile of these innovative, chiral ligands is covered below and highlights the impressive breadth of valuable transformations mediated by the various portfolio products. In many documented cases, specific ligands have displayed unprecedented selectivities in reactions that form, for instance, chiral centers in heteroatom-functionalized organic building blocks for drug synthesis.
Asymmetric Hydrogenation of the C=N Group
Burk and co-workers exploited the high activity of the Ethyl- DuPhos ligand via a powerful catalytic reductive amination process.4 The procedure exhibits general applicability in the reduction of a wide variety of N-arylhydrazones, yielding enantioselectivies for most substrates >90% (Scheme 3). Additionally, this Et-DuPhos-Rh catalyzed system displays exceptionally high chemoselectivies, yielding little or no reduction of unfunctionalized alkenes, alkynes, ketones, aldehydes, and imines in competition experiments. The synthetic utility of these asymmetric hydrazone reductions is enhanced by their facile reaction at ambient temperature with samarium diiodide to afford the corresponding chiral amines, which proceeds with no observable loss of optical purity.
Scheme 3
Catalytic Hydrogenation of Enamides
Burk also pioneered the asymmetric hydrogenation of various enamido olefins affording highly enantiopure d,g-unsaturated amino acid products.5 The (S,S)-Et-DuPhos-Rh catalyst system controls the reactivity of conjugated substrates with high regioselectivies as well. Under the standard hydrogenation conditions (S/C = 500, H2 pressures ranging from 60 to 90 psi, and 0.5–3 h), this catalyst gave less than 2% overreduction with all products isolated in better than 95% yield. The authors elaborated upon this outstanding catalyst reactivity by demonstrating a concise and highly selective synthesis of the natural product (–)-bulgecinine, preceeded by formation of the key chiral intermediate in 99% yield with 99.3% ee (Scheme 4).
Scheme 4
Highly Asymmetric Reductive Amidation
Burk and co-workers have also illustrated a rapid three-step process for reductive amidation, converting various ketones to the respective chiral amines with high enantioselectivity.6 Note that the transformation shown involves reacting the ketone with hydroxylamine followed by subsequent reduction with iron metal. This methodology benefits from the utilization of crude enamides isolated as crystalline solids in the direct asymmetric catalytic hydrogenation without prior purification. The conversion of 1-acetyladamantane to the novel amine compound occurs with >99% ee and in high yield via use of cationic Rh(I) catalyst 1 (Scheme 5).
Scheme 5
Preparation of Chiral Organics with C–O Stereogenic Centers
Neil Boaz utilized rhodium(I)-(R,R)-Me-DuPhos catalyst 1 to produce chiral alcohols via the asymmetric hydrogenation of enol esters (Scheme 6). Allylic alcohol derivatives are desirable organic building blocks in diversity-oriented synthesis, because the olefin can be further functionalized after the stereochemistry has been set in the hydrogenation.7 Under asymmetric hydrogenation conditions, the initially formed propargylic acetate was subsequently reduced to yield the Z-allylic acetate. Impressively, the enantioselectivity observed in this reaction was very high among the general substrate class 2a-c.
Scheme 6
Burk and co-workers have also designed highly effective catalysts for the asymmetric reduction of C=O bonds under hydrogenation conditions.8 In the case shown, the methodology proceeded via use of a chiral [Ru(II)Br2-(i-Pr-BPE)] complex, which was prepared by reacting [(cod)Ru(2-methylallyl)2] with the BPE ligand followed by treatment with methanolic HBr. A variety of esters were rapidly hydrogenated as mediated by this catalyst to the hydroxyl esters with very high enantioselectivities (>98%) ee for the alkylsubstituted substrates (Scheme 7).
Scheme 7
Preparation of Chiral Organics with C–C Stereogenic Centers
As a complement to the C–O bond-forming asymmetric synthesis, Burk and co-workers have performed highly enantioselective hydrogenations of unique substrates that are precursors to natural products. This methodology is highlighted by enantioselectivies approaching 100% and high efficiencies (S/C > 2500), and has allowed this research team to surmount the problems associated with the asymmetric synthesis of the drug Candoxitril (Scheme 8).9
Scheme 8
Novel Asymmetric [4+1] Cycloadditions
These highly practical enantioselective reactions have been accomplished by the Murkami research group, thus expanding the synthetic utility of these powerful chiral ligands.10 The [4+1] cycloaddition between vinylallenes and carbon monoxide affords complex cyclopentenone derivatives in a single step and with enantiopurities up to 95% (Scheme 9).
Scheme 9
Palladium-Catalyzed Asymmetric Phosphination
Glueck and co-workers have successfully developed a catalytic asymmetric phosphination reaction utilized in the production of P-chirogenic phosphines (Scheme 10).11 The fast conversions, moderate catalyst loadings, and superb enantioselectivies allow for the expeditious construction of enantioenriched phosphine building blocks. Metal-catalyzed routes to these valuable P-chiral ligands are rare making this methodology attractive as a viable route to synthesize other phosphine ligands.
Scheme 10
Catalytic Asymmetric Alkylation of N-Diphenylphosphinoylimines
Recently Charette has published a facile asymmetric synthesis of various a-chiral amines via the enantioselective addition of dialkylzinc reagents to N-phosphinoylimines.12 (R,R)-Me-DuPhos, in conjunction with a Cu(OTf)2 source, yielded exceptional enantiomeric excess for this reaction above 90% for all aromatic substrates (Scheme 11). The mild reaction conditions, reasonable temperatures, and use of various dialkylzinc reagents all contribute to the attractiveness of this methodology.
Scheme 11
Enantioselective Hydrogenation of Alkenes and Imines
The use of gold complexes to effect catalytic transformations is on the rise, with numerous reports of catalytically active gold species.13 Sanchez and co-workers have now reported the first example of a gold hydrogenation catalyst utilized in asymmetric transformations.14 The authors found that the bulkiest substrate, which incorporates a diethyl 2-naphthylidenesuccinate group, proceeds under the reaction conditions to afford the highest enantioselectivities due to reactant control (Scheme 12). Future plans in gold-mediated asymmetric hydrogenation involve substantial modifications to the ligand structure to provide higher levels of enantiocontrol.
Scheme 12
The First Catalytic Enantioselective Allylboration of Ketones
The Shibasaki research group has also championed use of the DuPhos ligand system for asymmetric catalysis.15 They have now reported the first general catalytic, enantioselective allylation reaction with ketones, which employs copper salts and a rareearth lanthanide additive. Impressively, a diverse array of aromatic, heteroaromatic, a,b-unsaturated, and aliphatic ketones are rapidly allylated at ambient temperature and under low catalyst loadings (Scheme 13). The enantioselectivities range from 67 to 92%, however, the reaction appears to be quite general for both the allylation and crotylboration reactions.
Scheme 13
back to top
Conclusion
The modular nature of these chiral phospholane ligands has allowed variation of the electronics and sterics around the metal upon complexation. These bidentate ligands perform highly enantioselective reactions to produce a wide array of compounds containing C–C, C–O, and C–N stereogenic centers. Sigma-Aldrich is your dedicated source for a broad spectrum of chiral building blocks that provide essential starting materials in the synthesis of complex organic molecules. Our growing portfolio of catalysis products, supplemented by the DuPhos/BPE family, strongly complements the existing Sigma-Aldrich chemical line and will accelerate your research success.
back to top
Materials
References
  1. Burk, M. J. Acc. Chem. Res. 2000, 33, 363.
  2. DuPhos and BPE ligands are sold in collaboration with Kanata Chemical Technologies and licensed from E. I. Dupont for the research market only.
  3. Burk, M. J. et al. J. Am. Chem. Soc. 1996, 118, 5142.
  4. Burk, M. J. et al. J. Am. Chem. Soc. 1992, 114, 6266.
  5. Burk, M. J. et al. J. Am. Chem. Soc. 1998, 120, 657.
  6. Burk, M. J. et al. J. Org. Chem. 1998, 63, 6084.
  7. Boaz, N. W. Tetrahedron Lett. 1998, 39, 5505.
  8. Burk, M. J. et al. J. Am. Chem. Soc. 1995, 117, 4423.
  9. Burk, M. J. et al. J. Org. Chem. 1999, 64, 3290.
  10. Murakami, M. et al. J. Am. Chem. Soc. 1995, 121, 4130.
  11. Moncarz, J. R. et al. J. Am. Chem. Soc. 2002, 124, 13356.
  12. Charette, A. B. et al. J. Am. Chem. Soc. 2003, 125, 1692.
  13. Dyker, G. Angew. Chem. Int. Ed. 2000, 39, 4237.
  14. Sanchez, F. et al. Chem. Commun. 2005, 3451.
  15. Shibasaki, M. et al. J. Am. Chem. Soc. 2004, 126, 8910.