Intermolecular [4+3]-Annulation
By: William Sommer, Aldrich ChemFiles 2009, 9.5, 4.
Aldrich ChemFiles 2009, 9.5, 4.
In 2008, Toste and coworkers developed a new method to synthesize azepines using a Au(III) catalyst.1 Following the work done by the group with sulfoxides showing that the allylgold intermediate can be induced to react as a nucleophile, Shapiro and Toste reacted various propargyl esters with various α,β-unsaturated imines (Scheme 8). It was noted that the highest yields were obtained with substrates containing electron-rich groups on the imine nitrogen. However, the reaction proved to be tolerant of variation at other positions. It was also noticed that electron-donating substituents on the N-aryl and β-aryl groups increase the rate of the cycloaddition.
Scheme 8.