Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ishikawa’s Reagent
Chemfiles Volume 5 Article 1
Ishikawa’s reagent (N,N-diethyl-1,1,2,3,3,3-hexafluoropropylamine) has also demonstrated utility in fluorination. In a recent example, Ishikawa’s reagent has been used in a clean and reproducible fluorination protocol on an intermediate in the total synthesis of 26-fluoroepothilone B(Scheme 14)1
Scheme 14
Ishikawa’s reagent also displays the ability to insert a fluoro(trifluoro-methyl)methylene moiety in unsaturated alcohols(Scheme 15).2
Scheme 15
back to top
References
  1. Koch, G. et al. Synlett. 2004, 693.
  2. Ogu, K-i. et al. Tetrahedron Lett. 1998, 39, 305.
back to top
Related Links
Categories