Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Leighton’s Chiral Allylation Reagents
By: Daniel Weibel, Chemfiles Volume 10 Article 3
Daniel Weibel, Ph.D. European Market Segment Manager, Chemistry
Daniel Weibel, Ph.D. European Market 
Segment Manager, Chemistry 
daniel.weibel@sial.com
The asymmetric allylation of carbonyl compounds remains one of the most important and fundamental addition reactions for the synthesis of optically active chiral building blocks homoallylic alcohols.
In 2002, Leighton and co-workers developed strained silacycle compounds as versatile reagents for the practical enantioselective allylation of aldehydes.1
A newly developed chiral auxiliary based on the cyclohexane-1,2- diamine scaffold successfully allylated a broad range of aldehydes highly enantioselectively (Scheme 1).2
Scheme 1. Leighton’s silacycle reagent for the enantioselective allylation of aldehydes
Scheme 1. Leighton’s silacycle reagent for the enantioselective allylation of aldehydes (704725)
The development of practical enantioselective syntheses of chiral amines is of great importance to synthetic organic and medicinal chemists. In 2003, Leighton and co-workers successfully used a pseudoephedrine-derived, five-membered-ring strained silacycle reagent for the enantioselective allylation of acylhydrazones (Scheme 2).3
Scheme 2. Practical, enantioselective allylation of acylhydrazones using strained silacycles
Scheme 2. Practical, enantioselective allylation of acylhydrazones using strained silacycles (706671)
The reaction scope of these silacycles was extended to a practical method for the enantioselective synthesis of tertiary carbinamines based on the addition of this chiral allylsilane reagent to a structurally diverse array of ketone-derived benzoylhydrazones (Scheme 3).4 While many methods for the synthesis of quaternary α-amino acids have been published, far fewer reports have dealt with the synthesis of tertiary carbinamines.
Scheme 3. Practical synthesis of tertiary carbinamines by enantioselective allylation of ketone-derived benzoylhydrazones
Scheme 3. Practical synthesis of tertiary carbinamines by enantioselective allylation of ketone-derived benzoylhydrazones (706671)
The free amines are easily accessed in good yields by subjecting the different hydrazones to reduction with SmI2 (409340).
back to top
Materials
References
  1. Leighton, J. L. Aldrichimica Acta 2010, 43, 3.
  2. Kinnaird, J. W. A.; Ng, P. Y.; Kubota, K.; Wang, X.; Leighton, J. L. J. Am. Chem. Soc. 2002, 124, 7920.
  3. Kubota, K.; Leighton, J. L. Angew. Chem., Int. Ed. 2003, 42, 946.
  4. Berger, R.; Rabbat, P. M. A.; Leighton, J. L. J. Am. Chem. Soc. 2003, 125, 9596.
  5. Berger, R.; Duff, K.; Leighton, J. L., J. Am. Chem. Soc. 2004, 126, 5686.