Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Mac-H: Asymmetric Hydrogenation in a Bottle
Chemfiles Volume 7 Article 5
MacMillan and co-workers have reported the combination of MacMillan Imidazolidinone OrganoCatalysts and Hantzsch esters to facilitate the first enantioselective organocatalytic hydride reduction of a, b-unsaturated aldehydes.1 Interestingly, isomerically pure E- and Z-olefin substrates will be reduced to the same enantiomer, allowing the researcher to use geometrically impure enals while maintaining excellent enantioselectivity. (S)-Mac-H is a convenient 6:1 formulation of the ethyl Hantzsch ester and OrganoCatalyst 1 for asymmetric reductions.
back to top
References
  1. Ouellet, S. G. et al. J. Am. Chem. Soc., 2005, 127, 32.