Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MacMillan Imidazolidinone OrganoCatalysts™
Aldrich ChemFiles 2006, 6.2, 9.
Aldrich ChemFiles 2006, 6.2, 9.
Metal-Free Asymmetric Catalysis
Product Highlights
  • Superior enantiocontrol in numerous transformations
  • High activities at low catalyst loadings
  • Extraordinary functional group tolerance
  • Asymmetric α-fluorination employed in natural product synthesis
MacMillan and co-workers have created chiral imidazolidinone organo-catalysts that function as the linchpin in a variety of directed enantioselective organic reactions, including the enamine-catalyzed α-chlorination and 1,3-dipolar cycloaddition of aldehydes. Sigma‑Aldrich is pleased to offer six imidazolidinone organocatalysts in our collaboration with Materia, Inc. that mediate rapid and enantiocontrolled C–F and C–H bond formation. In the former process, catalyst 1 was utilized in low (5 mol %) loadings in the first example of organocatalytic advanced enantioselective α-fluorination of aldehydes to afford a broad spectrum of highly enantioenriched alcohols.
back to top
Materials
References
    (a) MacMillan, D. W. et al. J. Am. Chem. Soc. 2000, 122, 9874. (b) MacMillan, D. W. et al. J. Am. Chem. Soc. 2005, 127, 8826.