MacMillan OrganoCatalysts™
Aldrich ChemFiles 2007, 7.3, 15.
Aldrich ChemFiles 2007, 7.3, 15.
Mac-H is a convenient formulation of the MacMillan Imidazolidinone OrganoCatalystTM and Hantzsch ester for asymmetric reductions, effectively serving as “asymmetric hydrogenation in a bottle.”
Mac-H
(683558)
MacMillan TiPSY Catalysts have been used in the first direct enantioselective organocatalytic reductive amination reaction.