MeO-BIPHEP
chemfiles volume 5 article 4
With Ru-complexes of this atropisomeric diphosphine ligand, b-keto esters are reduced to their corresponding b-hydroxy esters in high enantioselectivities. Selected results are shown in Scheme 13.
Scheme 13
Further enantioselective transformations of the MeO-BIPHEP ligands include Rh(I)-catalyzed asymmetric isomerizations of allylamines to enamines1 and Pd(0) complexes for cyclizations of hydroxy allylic carbonates.2
back to top 
References
  1. Schmid, R. et al. Helv. Chim. Acta 1991, 74, 370.
  2. Labrosse, J.-R. et al. Tetrahedron: Asymmetry 1999, 10, 1069.