Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MIDA Boronates for Classically Challenging Suzuki-Miyaura Cross-Couplings
By: Aaron Thornton, Chemfiles Volume 11 Article 1
The power of this slow-release concept has been further illustrated by utilizing various MIDA boronates of which the corresponding boronic acids have historically exhibited challenges with respect to either storage or use, including 2-heterocyclic, vinyl and cyclopropyl boronic acids. Because these organoboron species readily decompose through a variety of pathways, the efficiency with which their corresponding MIDA boronates may be coupled is particularily noteworthy (Table 1).
Table 1: Slow-release cross-coupling of MIDA boronates with historically challenging substrates.
back to top 
Related Links
Categories