Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Iodinated Building Blocks
By: Mark Redlich, Chemfiles Volume 10 Article 3
Iodine-containing substrates are valuable building blocks for a diverse array of synthetic methodologies. They have the ability to participate in various cross-coupling reactions for the generation of carbon–carbon, carbon–nitrogen, and carbon– oxygen bonds. Often iodinated substrates are preferred to the brominated analogs, due to cleaner reactions. They are also handy precursors for the formation of organolithium, organozinc, or organomagnesium reagents. Sigma-Aldrich is pleased to offer these useful building blocks for your research.