Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Olefin Metathesis
ChemFiles Volume 11 Article 1
Employing the second generation Grubbs catalyst (2 mol %) a variety of lypophillic substrates successfully undergo ring-closing or cross-metathesis in water at room temperature to produce high isolated yields of the desired products (Scheme 1). Reactions were conducted in 2.5% TPGS- 750-M/water, with yields equal to or slightly better than those performed using various other surfactant-water combinations.
Scheme 1 Selected olefin metathesis.
back to top Back to Top
Related Links