Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Pd(0) EnCat™ 30NP
Aldrich ChemFiles 2006, 6.5, 13.
Aldrich ChemFiles 2006, 6.5, 13.
Highly Versatile Catalyst for Chemoselective Hydrogenations
  • Safe and nonpyrophoric 
  • Trivial removal of catalyst from reactor 
  • Very low metal contamination of product 
  • Excellent catalyst recyclability 
  • Highly chemoselective under both hydrogenation & transfer hydrogenation conditions 
  • Excellent batch-to-batch reproducibility of catalyst activity
Pd(0) EnCat 30NP is a new microencapsulated hydrogenation catalyst offering key advantages over existing heterogeneous hydrogenation catalysts. Reductive transformations catalyzed by Pd(0) EnCat 30NP include aryl ketones, aldehydes, and epoxides to the corresponding alcohols; nitroarenes and aryl nitriles to the corresponding amines; alkenes and alkynes to the corresponding alkanes; and the debenzylation of aryl benzyl ethers.
back to top
Materials
References
  1. Bremeyer, N.; Ley, S. V.; Ramarao, C.; Shirley, I. M.; Smith, S. C. Synlett 2002, 1843. (2)(a) Yu, J.-Q.; Wu, H.-C.; Ramarao, C.; Spencer, J. B.; Ley, S. V. Chem. Commun. 2003, 678. (b) Ley, S. V.; Mitchell, C.; Pears, D.; Ramarao, C.; Yu, J.-Q.; Zhou, W. Org. Lett. 2003, 5, 4665.