Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
PhosphonicS™ Heterogeneous Metal Oxidation Catalysts
Chemfiles Volume 10 Article 2
        Dr. Josephine Nakhla
Dr. Josephine Nakhla
Product Manager
Email: josephine.nakhla@sial.com
Allylic and benzylic oxidations, alcohol oxidations, sulfoxidations, and epoxidations represent key synthetic transformations. Many traditional oxidants (chromium, manganesebased reagents, and peracids) are inherently toxic and often present difficulties in reaction work-up and purification, particularly from metal residues.
The development of new heterogeneous oxidation catalysts which would obviate the use of hazardous reagents such as those described above, would significantly advance the field. Under mild conditions in the presence of a re-oxidant, PhosphonicS heterogeneous oxidation catalysts cleanly, efficiently and selectively perform key oxidative transformations (Scheme 1) without the use of stoichiometric quantities of hazardous metalbased oxidants.1–4
Scheme 1:Versatility of PhosphonicS™ Heterogeneous Metal Oxidation Catalysts
Scheme 1:Versatility of PhosphonicS™ Heterogeneous Metal Oxidation Catalysts
Advantages of PhosphonicS Heterogeneous Metal Oxidation Catalysts:
  • Catalytic oxidants which circumvent the use of toxic stoichiometric oxidants
  • Simplified reaction work-up
  • Facile product isolation
  • Chemo and regioselective
back to top
N-Heterocyclic Carbene Organocatalysts
Organocatalysis has been an active field with over 2,000 publications in the past 10 years. Organocatalysis offers multiple advantages, such as simplified procedures, chemical waste reduction, obviating the need for a metal catalyst and subsequent product contamination, and inherent cost savings (since no metal catalyst is required). Scheidt and coworkers have developed a series of N-heterocyclic carbene organocatalysts for various transformations and have also developed chiral variants, which afford the desired products with good levels of enantioselectivity. The unusual but efficient conversion of α,β- unsaturated aldehydes to their saturated ester counterparts using a recently commercialized N-heterocyclic carbene as catalyst is depicted below (Scheme 2).
Scheme 2: NHC-catalyzed preparation of saturated esters from a,ß-unsaturated aldehydes
Scheme 2: NHC-catalyzed preparation of saturated esters from a,ß-unsaturated aldehydes
Subsequently, Scheidt and coworkers reported the use of a chiral triazole for the enantioselective addition of homoenolates to nitrones affording γ-amino esters (Scheme 3). The reaction scope was broad, proving amenable to both the use of electron-rich and electron-poor aldehydes. Using 20 mol % of the chiral triazole at –25 °C affords the desired products in good yields and selectivities.
Scheme 3: NHC-catalyzed enantioselective additions of enals to nitrones to afford disubstituted hydroxyl amines
Scheme 3: NHC-catalyzed enantioselective additions of enals to nitrones to afford disubstituted hydroxyl amines
back to top
Materials
References
  1. Al-Haq, N.; Sullivan, A. C.; Wilson, J. R. H. Tetrahedron Lett. 2003, 44, 769.
  2. Jurado-Gonzalez; M.; Sullivan, A. C.; Wilson, J. R. H. Tetrahedron Lett. 2003, 44, 4283.
  3. Jurado-Gonzalez; M.; Sullivan, A. C.; Wilson, J. R. H. Tetrahedron Lett. 2004, 45, 4465
  4. Al-Hashimi, M.; Roy, G.; Sullivan, A. C.; Wilson, J. R. H. Tetrahedron Lett. 2005, 46, 4365.
  5. Phillips, E.M.; Chan, A.; Scheidt, K.A. Aldrichimica Acta 2009, 42, 55.
  6. Chan, A.; Scheidt, K. A. Org. Lett. 2005, 7, 905.
  7. Phillips, E. M.; Reynolds, T. E.; Scheidt, K. A. J. Am. Chem. Soc. 2008, 130, 2416.