Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Piperidines
Aldrich ChemFiles 2007, 7.8, 7.
Aldrich ChemFiles 2007, 7.8, 7.
Piperidines have become increasingly popular building blocks in a vast array of synthetic protocols. 1-Boc-2-(aminomethyl)piperidine has been used in a post-Ugi carbonylation/intramolecular amidation approach to a series of eight-membered macrolactams with multiple points of diversity (Scheme 1). 1 On the other hand, the unprotected analogue has been employed in the synthesis of melanocortin 4 receptor antagonists (Scheme 2). These antagonists are potentially useful in the therapeutic treatment of involuntary weight loss. 2
Scheme 1(687154)
Scheme 2(656518)
Fluorinated piperidines are also showing increased interest in medicinal chemistry. Both 3- and 4-fluoropiperidine were used in the synthesis of selective dipeptidyl peptidase II (DPP II) inhibitors (Scheme 3). The 3-fluoro analogue in particular demonstrated improved selectivity, without sacrificing potency. 3
Scheme 3(680168)(665606)
back to top
Materials
References
  1. Vasudevan, A.; Verzal, M. K. Tetrahedron Lett. 2005, 46, 1697.
  2. Marsilje, T. H. et al. Bioorg. Med. Chem. Lett. 2004, 14, 3721.
  3. Senten, K. et al. J. Med. Chem. 2004, 47, 2906.