Pt(0) EnCat™ 40
Chemfiles Volume 7 Supplement 1
Pt(0) EnCat 40 is a versatile hydrogenation catalyst demonstrating high chemoselectivity and improved safety profile over platinum on carbon allowing easier, faster and cleaner processes to be developed. Isolating the active platinum within the polymer matrix has produced an easily handled and safer alternative to Pt/C.
Cleaner products
very low levels of Pt in crude product
Cleaner waste streams
reduced metal losses in EnCat process (typically <0.005% Pt loss)
Faster, efficient processes
EnCat beads filter easily
No plant contamination
platinum metal remains entrapped within the beads
High selectivity
in hydrogenation reactions
Process intensification
can be used in batch & flow processes
Improved safety profile
initial tests show no tendency for activated catalyst to ignite upon exposure to air
back to top
Related Links
Categories