Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
(R)-(+)- and (S)-(–)-Stericol® New, Effective Chiral Auxiliaries
By: Mark Redlich, Chemfiles Volume 9 Article 3
The introduction of chirality to organic molecules through the use of auxiliaries is a basic strategy in organic chemistry. Chiral benzylic alcohols are of particular interest because they often afford useful levels of stereoselection and are generally easy to cleave.
Preparation of Dichlorocyclobutanones
Preparation of Dichlorocyclobutanones
Asymmetric Oxidation of Benzenesulfenates
Asymmetric Oxidation of Benzenesulfenates
Preparation of β,γ-Unsaturated Silyl Sulfinates
Preparation of β,γ-Unsaturated Silyl Sulfinates
The enantiomers of 1-(2,4,6-triisopropyl)phenylethanol ((R)- and (S)-Stericol®) are especially beneficial as chiral auxiliaries, providing high diasteromeric ratios in [2+2] cycloadditions,1 oxidations,2and carbon-carbon bond forming reactions.3
back to top 
References
  1. (a) de Azevedo, M. B. M.; Greene, A. E. J. Org. Chem. 1995, 60, 4940. (b) Kanazawa, A. et al. J. Chem. Soc. Perkin Trans. 1997,1911. (c) Nebois, P.; Greene, A. E. J. Org. Chem. 1996, 61, 5210. (d) Ceccon, J. et al. Synlett 2005, 1413. (e) Roche, C. et al. Org. Lett. 2003, 5, 1741. (f) Muniz, M. N. et al. Synlett 2005, 1328. (g) Roche, C. et al. J. Org. Chem. 2005, 70, 8352. (h) Ceccon, J. et al. Org. Lett. 2006, 8, 4739.
  2. Hamel, M.; Grach, G.; Abrunhosa, I.; Gulea, M.; Masson, S.; Vazeux, M.; Drabowicz, J.; Mikolajczyk, M. Tetrahedron: Asymmetry 2005, 16, 3406.
  3. (a) Narkevitch, V. et al. Angew. Chem., Int. Ed. 2000, 39, 1806. (b) Bouchez, L. C. et al. Tetrahedron 2005, 61, 11473. (c) Narkevitch, V. et al. J. Org. Chem. 2001, 66, 5080.