Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Reetz Ligands
By: William Sommer and Daniel Weibel, Aldrich ChemFiles 2008, 8.2, 54.
Aldrich ChemFiles 2008, 8.2, 54.
In 1998, Reetz et al. reported the synthesis of a new generation of ligand for enantioselective, catalyzed hydrogenation. Based on BINOL-derived diphosphonite molecules, these ligands showed good enantioselectivities for the asymmetric hydrogenation of terminal alkenes, ketones, and β-keto esters,1,2 and the asymmetric conjugate addition of arylboronic acid derivatives to α,β-unsaturated carbonyls.3
back to top
Enantioselective Hydrogenation of Olefins
Reetz et al. reported the hydrogenation of two different olefins with Rh(cod)BF4 and R,R-Reetz F-Diphosphonite. Outstanding yields and selectivity were reported (Scheme 1).4 Used with a RuCl2(p-cymene)2 complex, the R,R-Reetz X-Diphosphonite converts a variety of ketones into secondary alcohols with yields and ee's up to 100% and 98%, respectively (Scheme 2).1
Scheme 1
Scheme 1
Scheme 2
Scheme 2
back to top
Materials
References
  1. Reetz, M. T. et al. J. Am. Chem. Soc. 2006, 128, 1044.
  2. Reetz, M. T. et al. Adv. Synth. Catal. 2006, 348, 1157.
  3. Reetz, M. T. et al. Org. Lett. 2001, 3, 4083.
  4. Reetz, M. T. et al. Chem. Commun. 1998, 2077.