Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Resins for the Boc-/Bzl-Synthesis of C-terminal Modified Peptides
Chemfiles Volume 3 Article 4
The 4-nitrobenzophenone oxime resin has been used extensively for the preparation of Boc-/Bzl-protected peptide fragments.(1-3) Various nucleophiles have been described to cleave the peptide resin linkage to release different C-terminal modified peptides.(4-6) Cyclization cleavage provides cyclic peptides.(7-9)
73083 4-Nitrobenzophenone oxime, polymer-bound, Kaiser oxime resin
  polymer matrix: polystyrene cross-linked with 1% DVB
  capacity: ~ 1.1 mmol/g resin particle size 100–200 mesh 1 g, 5 g
back to top 
Materials
References
  1. DeGrado W. F.; Kaiser E. T. J. Org. Chem. 1980, 45, 1295.
  2. DeGrado W. F.; Kaiser E. T. J. Org. Chem. 1982, 47, 3258.
  3. Kaiser E.T. et al. Science 1989, 241, 187.
  4. Hendrix, J. C. et al. J. Org. Chem. 1992, 57, 3414.
  5. Pichette, A. et al. Tetrahedron Lett. 1997, 38, 1279.
  6. Sasaki, T. et al. J. Org. Chem. 1991, 56, 3159.
  7. Oesapay, G.; Taylor, J. W. J. Am. Chem. Soc. 1990, 112, 6046.
  8. Oesapay, G. et al. Tetrahedron Lett. 1990, 31, 6121.
  9. Nishino, N. et al. J. Chem. Soc., Chem. Commun. 1992, 180.
back to top 
Related Links
Categories