Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Synthesis of Brush-like Polymers via ROMP
By: William Sommer, Aldrich ChemFiles 2009, 9.6, 8.
Aldrich ChemFiles 2009, 9.6, 8.
Recently, Cheng and coworkers reported the synthesis of brush-like polymers, by which the main polymer chain was prepared using ROMP followed by polymerization of peptide units on the amine side chain with N-carboxyanhydrides. These new materials may extend the application of brush-like polymers to include new ranges of properties. In this report, the authors use the Grubbs catalyst (3rd Generation) to polymerize the norbornene monomers. Excellent polydispersity of the polymer was reported with PDI ranges of 1.03 to 1.15 (Scheme 1).1
Scheme 1
back to top Back to Top
Materials
References
  1. Lu, H. et al. J. Am. Chem. Soc.,2009, 131, 13582.
back to top Back to Top
Related Links