Anhydrous and Air-Sensitive Reagents
By: Todd Halkoski, Chemfiles Volume 10 Article 2
           Todd Halkoski
Todd Halkoski
Market Segment Manager
Email:todd.halkoski@sial.com
The Ideal Solution for Anhydrous and Air-Sensitive Reagents
As the leader in air-sensitive chemistry, Sigma-Aldrich carries by far the largest breadth of anhydrous solvents and air-sensitive reagents to service the research market. In the late 1970’s, Dr. Clint Lane developed the Sure/Seal bottle to protect sensitive reagents from oxygen and moisture. This not only maintained product integrity, but also facilitated the use of these products.
Our traditional Sure/Seal is now undergoing major improvements. During the upcoming months, you will begin experiencing the value of these new design improvements.
back to top
New Crimp Cap Design
  • Increased puncture area by 4x to accommodate multiple punctures
  • Crimp cap helps maintain product quality by providing an air-tight system
New Aldrich Sure/Seal
New Aldrich Sure/Seal
Previous Aldrich Sure/Seal
Previous Aldrich Sure/Seal
back to top
New Plug Style
  • Maximimizes surface area contact with the bottle to prevent moisture and oxygen entry
  • More than 50% thicker than competing brands
New plug style
back to top
New Elastomer Liner
Provides outstanding resealing properties
  • Secondary resin layer ensures resistance to chemicals
  • Outperforms competing seals with respect to moisture uptake
New Aldrich Sure/Seal System
Unpunctured
Unpunctured
4 Punctures
4 Punctures
Competitor A System
Unpunctured
Unpunctured
4 Punctures
4 Punctures
back to top
Aldrich Sure/Seal Testing Results
To illustrate the effectiveness of the new Aldrich Sure/Seal, our design was tested against another supplier, which also sells similar anhydrous solvents. Specifically, we compared their performance based upon:
  1. The liners ability to seal and maintain product quality after repeated punctures. Specifically, we measured the rate of moisture gain by Karl Fischer analysis over a five-week testing period.
  2. The amount of oxygen allowed to permeate into the bottle over a period of time. The Oxygen Transmission Rate (OTR) testing demonstrates the cap and liners ability to maintain an airtight seal when both unpunctured and punctured.
  3. The integrity of the liner to hold its shape and continue to perform in a laboratory setting after receiving numerous punctures without risk of liner degradation, deformation or catastrophic failure.
back to top
Karl Fischer Analysis
Utilizes a coulometric titration method to measure the rate of moisture uptake. To determine the effectiveness of the liner’s seals and closure design, a five-week study was conducted.
Karl Fischer Analysis
Comprehensive testing confirms the New Aldrich Sure/Seal outperforms other products on the market by maintaining low water absorption even after multiple punctures.
back to top
Oxygen Transmission Rate Testing
To demonstrate the effectiveness of our new Aldrich Sure/Seal, we evaluated it against another supplier to see how each performed when tested for the amount of oxygen allowed to permeate into the bottle.
Oxygen Transmission Rate Comparison
* Liners were punctured 4 times with an 18-gauge needle, and overlaps were torqued to 40 in lbs.
OTR testing demonstrates the new Aldrich Sure/Seal sustains product quality better than the competition.
back to top
Liner Integrity
When using air-sensitive solvents and reagents, liners should not only seal to maintain product quality, but also consistently hold form after repeated puncture. The New Aldrich Sure/Seal gives you the assurance of safety and a quality product.
Toluene Testing—5 Weeks (20 punctures)
Competitor A
Competitor A
New Sure/Seal System
New Sure/Seal System