Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ultra-Fast Initiating Ruthenium Catalysts for Low-Temperature Metathesis
By: Dr. Josephine Nakhla, Chemfiles Volume 10 Article 1
Traditional Grubbs catalyst systems are five-coordinate ruthenium complexes containing two neutral ligands, one of which is typically a phosphine, or in the case of the Hoveyda-Grubbs catalysts, a styrenyl ether. Ligand dissociation is required to provide the active catalyst, but is slow at low temperatures and, therefore, traditional Grubbs catalysts suffer from decreased reactivity at low temperatures. The Piers group developed preformed 4-coordinate cationic complexes that do not require ligand dissociation prior to reaction, allowing for efficient metathesis at lower temperatures. The reaction progress was examined for the cyclization of diethyldiallylmalonate to provide the corresponding cyclopentene derivative using both the Piers catalyst and the Grubbs catalyst at 0 °C (Scheme 2). At this temperature, the Grubbs catalyst (2nd Generation) was found to be a weak initiator and, after 4 hours, the reaction had progressed only to 25% completion, while the cationic complex had progressed to >90% completion after 2 hours. The initiation rate of the cationic catalyst at 0 °C was found to be comparable to the initiation rate of the Grubbs catalyst (2nd Generation) at 35 °C. In a subsequent study, Piers and coworkers examined the intermediates in an olefin metathesis reaction by NMR at –50 °C. This was the first direct observation of a ruthenacyclobutane intermediate and provided evidence for a symmetrical intermediate. It also illustrated the stabilizing effect of the N-heterocyclic carbene ligand on the Ru(IV) species.1
Scheme 2: Fast initiating ruthenium catalysts for low-temperature metathesis.
Scheme 2: Fast initiating ruthenium catalysts for low-temperature metathesis.
back to top
Materials
References
  1. (a) Romero, P. E. et al. Angew. Chem. Int. Ed. 2004, 43, 6161. (b) Romero, P. E.; Piers, W. E. J. Am. Chem. Soc. 2005, 127, 5032.
  2. Schrock, R. R. Chem. Rev. 2002, 102, 145.
  3. (a) Fürstner, A. et al. Chem. Commun. 2005, 2307. (b) Schrock, R. R. et al. Adv. Synth. Catal. 2007, 349, 55.
  4. Bindl, M. et al. J. Am. Chem. Soc. 2009, 131, 9468.