Potential Application for 2-Methoxythiophenol, FG
Available in Halal
At low levels this ingredient’s smoky phenolic notes find application in a variety of smoke flavors and savory meat modifiers. This product also finds use in vanilla top notes and grape wine musks making it a versatile ingredient.
Key Organoleptic Characteristics   Sulfurous, sweet, spicy, smoky, savory vegetative, meaty, slight vanilla and grape
Potential Applications Smoky, savory meat flavors with vanilla or grape-like top notes
Natural Occurrence Not yet found in nature
Aroma Characteristics @1.0% Ethanol. Sulfurous, sweet, spicy, smoky with savory vegetative and meaty nuances. Slight vanilla and grape-like notes.
Taste Characteristics @<1ppm. Sweet sulfurous wood smoke and vanilla with savory earthy nuances of potato, radish and cereal grains.
Product Number W415901