Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Potential Application for Vanillyl Butyl Ether
Available in Natural (US) & Kosher
Vanillyl butyl ether is useful in preparing Jamaican ginger beer flavors where the ginger after taste heat is a must. It could also be used for alcohol flavors where the throat burn would be useful.
Key Organoleptic Characteristics   Sweet, vanillin, creamy with a pronounced tongue bite and lingering peppery burn
Potential Applications Pepper additives; confections and chewing gum flavors for trigeminal effects; ginger spice blends
Natural Occurrence Not yet found in nature
Aroma Characteristics @10 % in ethanol. Heavy, sweet, creamy vanillin, with a slight spicy coumarin like nuance
Taste Characteristics @10-15 ppm. Initially creamy vanillin and coumarin-like, then there is a perceived tongue bite followed by an interesting and lingering peppery burn
Product Number W379601