Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Palladium-Catalyzed N-Arylation of 2-Aminothiazoles
Reaction Scheme
Abstract
The chemoselective and complementary Pd- and Cu-catalyzed N-arylation of 2-aminobenzimidazoles is described. Selective N-arylation of the amino group was achieved with a Pd-catalyzed method, while selective N-arylation of azole nitrogen was achieved with a Cu-catalyzed procedure
Substrates
Procedure
An oven-dried vial was equipped with a magnetic stir bar and charged with Pd2(dba)3 (2.3 mg, 0.0025 mmol) and Me4tBuXPhos (2.4 mg, 0.005 mmol). The vial was sealed with a screw-cap septum, and then evacuated and backfilled with argon (this process was repeated a total of 3 times). Anhydrous t-BuOH (0.3 mL) was added via syringe and the resulting dark purple mixture was stirred at 120 �C for 3 min, at this point the color of the mixture turned to red-brown. A second oven-dried vial, which was equipped with stir bar, was charged with aminoazole (1.1 mmol) and K3PO4 (318 mg, 1.5 mmol) (aryl halides that were solid at room temperature were added at this point). The vial was sealed with a screw-cap septum, and then evacuated and backfilled with argon (4 psi) (this process was repeated a total of 3 times). Aryl halide (1.0 mmol) and the pre-heated catalyst solution (0.12-0.3 mL, 0.2-0.5 mol% Pd) were added by syringe to the second vial, followed by addition of t-BuOH (1.5 mL). The reaction mixture was heated at 120 �C for 5 h. The reaction mixture was cooled to room temperature, diluted with EtOAc, washed with brine and dried over MgSO4. Silica gel (1.0 g) was added to the mixture and the mixture was concentrated in vacuo. The Silica gel-adsorbed product was purified via flash chromatography