Pd-Catalyzed Nucleophilic Fluorination of Aryl Bromides
Reaction Scheme
Abstract
The first palladium-catalyzed amidation of five-membered heterocyclic bromides with multiple heteroatoms was achieved using the Pd/1�catalyst system. N-Arylated imidazoles, pyrazoles, thiazoles, pyrroles, and thiophenes were synthesized in moderate to excellent yield. Experimental results and DFT calculations point to the need for an electron-rich and sterically demanding biaryl phosphine ligand to promote these difficult reactions.
Substrates
Procedure
General procedure: An oven-dried test tube was equipped with a magnetic stir bar and charged with [{(allyl)PdCl}2] (0.75 mol %), AdBrettPhos (3.0 mol %), Cs2CO3 (2 mmol), and amide (2 mmol ; the heteroaryl bromide (1 mmol), if solid, is added at this point). The test tube was sealed with a screw-cap septum, and then evacuated and backfilled with argon (this process was repeated a total of 3 times). 2-Methyl-2- butanol (2 mL) and heteroaryl bromide (1 mmol) were then added by syringe. The reaction mixture was heated at 90�C for 21h. The reaction mixture was cooled to room temperature, diluted with EtOAc, washed with a saturated solution of sodium bicarbonate, dried over Na2SO4, concentrated in vacuo, and purified by flash chromatography on silica gel (see the Supporting Information for details) to give pure products.