Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Grubbs Group - Professor Product Portal
Professor Product Portal Index
Professor Phil S Baran Professor Robert H. Grubbs
Research in the Grubbs group has centered on the development and application of a suite of highly active, selective, and bench stable ruthenium alkylidene complexes capable of catalyzing versatile olefin metatheses. Most notably, access to monodisperse polymers and macrocyclic/linear small molecules has been enabled through the use of these precatalysts. The ligands on ruthenium include, but are not limited to, neutral phosphines, N-heterocyclic carbenes, and pyridyl moieties, which allow for manipulations of initiation and/or propagation activity, stability, and selectivity. Similar accomplishments have been achieved with anionic ligand variations. Traditionally, olefin metathesis predominantly forms the thermodynamically favored (E)-olefin. Recent developments, however, which make use of cyclometallated N-heterocyclic carbene ruthenium alkylidene complexes, have unveiled a catalyst that forms (Z)-olefins almost exclusively. Advances in metathesis catalysts in the Grubbs group have had ubiquitous applications in the fields of energy, materials, pharmaceuticals, and natural product total synthesis.
For applicable product information, please click below:
For recent articles from the Grubbs Laboratory:
Products available at Aldrich from the Grubbs Laboratory
Adam M Johns, Tonia S Ahmed, Bradford W Jackson, Robert H Grubbs, Richard L Pederson
Organic Letters 2016-02-19
The first kinetically controlled, highly trans-selective (>98%) olefin cross-metathesis reaction is demonstrated using Ru-based catalysts. Reactions with either trans or cis olefins afford products with highly trans or cis stereochemistry, respectively. This E-selective olefin cross-metathesis is shown to occur between two trans...Read More
Peter K Dornan, Zachary K Wickens, Robert H Grubbs
Angewandte Chemie. International edition in English 2015-06-08
A stereoselective synthesis of anti-1,2-diols has been developed using a multitasking Ru catalyst in an assisted tandem catalysis protocol. A cyclometalated Ru complex catalyzes first a Z-selective cross-metathesis of two terminal olefins, followed by a stereospecific dihydroxylation. Both steps are catalyzed by Ru, as the Ru co...Read More
Keary M Engle, Gang Lu, Shao-Xiong Luo, Lawrence M Henling, Michael K Takase, Peng Liu, K N Houk, Robert H Grubbs
Journal of the American Chemical Society 2015-05-06
A series of second-generation ruthenium olefin metathesis catalysts was investigated using a combination of reaction kinetics, X-ray crystallography, NMR spectroscopy, and DFT calculations in order to determine the relationship between the structure of the chelating o-alkoxybenzylidene and the observed initiation rate. Included ...Read More
Myles B Herbert, Robert H Grubbs
Angewandte Chemie. International edition in English 2015-04-20
Olefin cross metathesis is a particularly powerful transformation that has been exploited extensively for the formation of complex products. Until recently, however, constructing Z-olefins using this methodology was not possible. With the discovery and development of three families of ruthenium-based Z-selective catalysts, the f...Read More
Vanessa M Marx, Alexandra H Sullivan, Mohand Melaimi, Scott C Virgil, Benjamin K Keitz, David S Weinberger, Guy Bertrand, Robert H Grubbs
Angewandte Chemie. International edition in English 2015-02-02
An expanded family of ruthenium-based metathesis catalysts bearing cyclic alkyl amino carbene (CAAC) ligands was prepared. These catalysts exhibited exceptional activity in the ethenolysis of the seed-oil derivative methyl oleate. In many cases, catalyst turnover numbers (TONs) of more than 100,000 were achieved, at a catalyst l...Read More