Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Zhang Group - Professor Product Portal
Professor Product Portal Index
Professor Liming Zhang
The major research thrust in Professor Liming Zhang’s group is homogeneous gold catalysis ranging from discovery of novel reactivities, design of new ligands, development of versatile synthetic methods and application in natural product synthesis. The pioneer contributions of his research group to the rapid rise of gold chemistry include tandem transformations of propargylic carboxylates, cycloaddition reactions with gold-containing all-carbon 1,n-dipoles, cross coupling reactions via Au(I)/Au(III) catalysis, gold vinylidene chemistry, and catalyzed alkyne oxidation. The last topic, especially when using an external oxidant, offers facile access to synthetically highly versatile α-oxo gold carbenes from readily available alkynes in an operationally safe manner. In contrast, the conventional approach to generating these intermediates requires the use of hazardous and potentially explosive diazo ketones, which in addition are often difficult and/or inefficient to synthesize. His group has introduced into the field several new and effective gold(I) complexes with bulky and electron-rich phosphine ligands (including phosphines developed by Buchwald and Stradiotto), and is currently working on the development of novel ligands for highly versatile gold catalysis.
Zhang Group website
For applicable product information, please click below:
For recent articles from the Zhang Laboratory
Products available at Aldrich from the Zhang Laboratory
Yanzhao Wang, Kegong Ji, Sylvester Lan, Liming Zhang
Angewandte Chemie. International edition in English 2012-02-20
Longwu Ye, Weimin He, Liming Zhang
Angewandte Chemie. International edition in English 2011-03-28