High-Tech Carbons
Material Matters 2009, 4.4, 110.
Material Matters 2009, 4.4, 110.
Engineered for Applications on This World and Beyond
The Sigma-Aldrich® commitment to carbon materials research and product development spans more than two decades. Understanding how thermodynamic and kinetic properties affect performance characteristics of both existing and novel carbons is our primary focus. The knowledge gained from this fundamental research has led to innovative new carbons and subsequent advances in the performance of many others. Today, we offer over 45 different carbons ranging in particle size from <0.2 to 841 μm and surface areas from 5 to 1,500 m2/g.
The unique and valuable characteristics of our carbon materials warranted their inclusion in experiments onboard the 1995 Galileo Mission to Jupiter, the 2005 Cassini-Huygens Missions to Saturn’s largest moon Titan, and the 2007 Phoenix Mission to Mars.
An important area of rapidly growing interest is the use of mesoporous and high surface area carbons in supercapacitor research. A list of selected Sigma-Aldrich carbon materials and their physical characteristics are shown in the Table 1. For a complete list of our materials for energy applications, visit sigma-aldrich.com/renewable.
  Pore Volume (cc/g)   Particle Size  
Prod. No. Carbon Surface
Area (m2/g)
Micro Meso Macro Pore
Diameter (Å)
Mesh μm Pkg
Size (g)
699624-5G Mesoporous carbon, graphitized 70       137   <0.5 5
699640-5G Mesoporous carbon 203 - 0.49 - 96.3 - 40-50 5
699632-5G Mesoporous carbon, purified 214 0.06 0.28 - 63.9 - <0.2 5
11051-U Carbopack™ Z 220 - 1.73 - 255 60/80 177-250 10
10439-U Carbopack X 240 - 0.62 - 100 120/400 37-125 50
10264 Carboxen™ 564 400 0.24 0.13 0.14 6-9 20/45 354-841 10
10269 Carboxen 569 485 0.20 0.14 0.10 5-8 20/45 354-841 10
10263 Carboxen 563 510 0.24 0.15 0.24 7-10 20/45 354-841 10
10184 Carbosieve™ S-III 975 0.35 0.04 - 4-11 60/80 177-250 10
10471 Carboxen 1003 1,000 0.38 0.26 0.28 5-8 40/60 250-400 10
10190-U Carbosieve S-II 1,059 0.45 0.01 - 6-15 80/100 149-177 10
11072-U Carboxen 572 1,100 0.41 0.19 0.24 10-12 20/45 354-841 10
10199 Carbosieve G 1,160 0.49 0.02 - 6-15 80/100 149-177 5
11052-U Carboxen 1000 1,200 0.44 0.16 0.25 10-12 80/100 149-177 100
Table 1. High surface area carbons
back to top Back to Top