Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Aligned Multi-Walled Carbon Nanotubes for High-Sensitivity Gas Sensors
By: Joe Porwoll, Material Matters 2007, 2.4, 3.
Aligned Multi-Walled Carbon Nanotubes (MWCNTs)
Dr. Karl Gross, of Hy-Energy LLC, kindly suggested that high-quality carbon nanotube arrays are needed for research and development of high-sensitivity gas sensors. We are pleased to offer highly aligned arrays of multiwall carbon nanotubes (MWCNTs) as new products in our catalog. These arrays, available on silicone (Aldrich Product No. 687804) or copper (Aldrich Product No. 687812) substrates, can provide a unique platform for a wide range of materials research and development, including gas adsorption sensor and sensor substrates,1 catalysts, electron emission sources,2 and battery and capacitor development.3
Carbon nanotube array, multi-walled, vertically aligned, on silicon wafer substrate
687804-1EA
Carbon nanotube array, multi-walled, vertically aligned, on copper substrate
687812-1EA
CNT array (black square) in the semiconductor grade package. Inset shows SEM image of the vertically aligned MWCNTs.
back to top
Material Matters 2007, 2.4, 3.
Features:
  • 99.9% as MWCNT
  • CNT diameters are 100 nm ± 10 nm
  • CNT lengths are 30 μm ± 3 μm
  • Array density ~2 x 109 MWCNT/cm2
  • Array dimensions 1 sq. cm
  • Grown by plasma-enhanced CVD (PECVD)
  • Grown with the nickel catalyst tip intact
  • Packaged in a clean room; stored, and shipped in semiconductor grade packaging.
back to top
Materials
References
  1. Collins, P.G., Bradley, K. Ishigami, M. Zettl, A., Science, 2000, 287, 1801.
  2. Bonard, J. M., Stockli, T., Maier, F., De Herr, W. A., Chatelain, A., Ugarte, D., Salvetat, J. P., Forro, L., Physical Review Letters, 1998, 81, 1441.
  3. Frackowiak, E., Gautier, S., Gaucher, H., Bonnamy, S., Beguin, F., Carbon, 1999, 37, 61